missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human XRN2 Partial ORF (NP_036387.2, 516 a.a. - 615 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
auf Anfrage
Spezifikation
Name | XRN2 (Human) Recombinant Protein (Q01) |
---|---|
Rekombinant | Yes |
Molekulargewicht | 36.74kDa |
Lagerungsbedingungen | Store at -80°C. Aliquot to avoid repeated freezing and thawing. |
Beschreibung
This gene shares similarity with the mouse Dhm1 and the yeast dhp1 gene. The yeast gene is involved in homologous recombination and RNA metabolism, such as RNA synthesis and RNA trafficking. Complementation studies show that Dhm1 has a similar function in mouse as dhp1. The function of the human gene has not yet been determined. Transcript variants encoding different isoforms have been noted for this gene; however, their full-length nature is not known. [provided by RefSeq]
Sequence: WKQRYYKNKFDVDAADEKFRRKVVQSYVEGLCWVLRYYYQGCASWKWYYPFHYAPFASDFEGIADMPSDFEKGTKPFKPLEQLMGVFPAASGNFLPPSWRSpezifikation
XRN2 (Human) Recombinant Protein (Q01) | |
Yes | |
36.74kDa | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. |