Learn More
Abnova™ Human WIPI2 Full-length ORF (NP_056425.1, 1 a.a. - 454 a.a.) Recombinant Protein with GST-tag at N-terminal
Beschreibung
WD40 repeat proteins are key components of many essential biologic functions. They regulate the assembly of multiprotein complexes by presenting a beta-propeller platform for simultaneous and reversible protein-protein interactions. Members of the WIPI subfamily of WD40 repeat proteins, such as WIPI2, have a 7-bladed propeller structure and contain a conserved motif for interaction with phospholipids (Proikas-Cezanne et al., 2004 [PubMed 15602573]).[supplied by OMIM]
Spezifikation
Spezifikation
Zugriffsnummer | NP_056425.1 |
Zur Verwendung mit (Anwendung) | Antibody Production, ELISA, Protein Array, Western Blot |
Zusammensetzung | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gen-ID (Entrez) | 26100 |
Molekulargewicht | 75.8kDa |
Name | WIPI2 (Human) Recombinant Protein (P01) |
Reinigungsverfahren | Glutathione Sepharose 4 Fast Flow |
Qualitätskontrollen | 12.5% SDS-PAGE Stained with Coomassie Blue. |
Menge | 25 μg |
Immunogen | MNLASQSGEAGAGQLLFANFNQDNTEVKGASRAAGLGRRAVVWSLAVGSKSGYKFFSLSSVDKLEQIYECTDTEDVCIVERLFSSSLVAIVSLKAPRKLKVCHFKKGTEICNYSYSNTILAVKLNRQRLIVCLEESLYIHNIRDMKVLHTIRETPPNPAGLCALSINNDNCYLAYPGSATIGEVQVFDTINLRAANMIPAHDSPLAALAFDASGTKLATASEKGTVIRVFSIPEGQKLFEFRRGVKRCVSICSLAFSMDGMFLSASSNTETVHIFKLETVKEKPPEEPTTWTGYFGKVLMASTSYLPSQVTEMFNQGRAFATVRLPFCGHKNICSLATIQKIPRLLVGAADGYLYMYNLDPQEGGECALMKQHRLDGSLETTNEILDSASHDCPLVTQTYGAAAGKGTYVPSSPTRLAYTDDLGAVGGACLEDEASALRLDEDSEHPPMILRTD |
Mehr anzeigen |
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.