missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human WDR89 Full-length ORF (NP_001008726.1, 1 a.a. - 387 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marke:  Abnova™ H00112840-P01.25ug

Additional Details : Gewicht : 0.00010kg

 Lernen Sie weitere Varianten dieses

Artikelnummer. 16125519

  • 574.00 CHF / 25 Mikrogramm
Verfügbar ab: 17-01-2024
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

Sequence: MEKIEEQFANLHIVKCSLGTKEPTYLLGIDTSKTVQAGKENLVAVLCSNGSIRIYDKERLNVLREFSGYPGLLNGVRFANSCDSVYSACTDGTVKCWDARVAREKPVQLFKGYPSNIFISFDINCNDHIICAGTEKVDDDALLVFWDARMNSQNLSTTKDSLGAYSETHSDDVTQVRFHPSNPNMVVSGSSDGLVNVFDINIDNEEDALVTTCNSISSVSCIGWSGKGYKQIYCMTHDEGFYWWDLNHLDTDEPVTRLNIQDVREVVNMKEDALDYLIGGLYHEKTDTLHVIGGTNKGRIHLMNCSMSGLTHVTSLQGGHAATVRSFCWNVQDDSLLTGGEDAQLLLWKPGAIEKTFTKKESMKIASSVHQRVRVHSNDSYKRRKKQ
Spezifikation

Spezifikation

NP_001008726.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
69.6kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
C14orf150/MGC9907/MSTP050
WDR89
Recombinant
wheat germ expression system
Antibody Production, ELISA, Protein Array, Western Blot
112840
WDR89 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MEKIEEQFANLHIVKCSLGTKEPTYLLGIDTSKTVQAGKENLVAVLCSNGSIRIYDKERLNVLREFSGYPGLLNGVRFANSCDSVYSACTDGTVKCWDARVAREKPVQLFKGYPSNIFISFDINCNDHIICAGTEKVDDDALLVFWDARMNSQNLSTTKDSLGAYSETHSDDVTQVRFHPSNPNMVVSGSSDGLVNVFDINIDNEEDALVTTCNSISSVSCIGWSGKGYKQIYCMTHDEGFYWWDLNHLDTDEPVTRLNIQDVREVVNMKEDALDYLIGGLYHEKTDTLHVIGGTNKGRIHLMNCSMSGLTHVTSLQGGHAATVRSFCWNVQDDSLLTGGEDAQLLLWKPGAIEKTFTKKESMKIASSVHQRVRVHSNDSYKRRKKQ
RUO
WDR89
Wheat Germ (in vitro)
GST
Liquid
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote