Learn More
Abnova™ Human WASF2 Full-length ORF (NP_008921.1, 1 a.a. - 498 a.a.) Recombinant Protein with GST-tag at N-terminal
Beschreibung
This gene encodes a member of the Wiskott-Aldrich syndrome protein family. The gene product is a protein that forms a multiprotein complex that links receptor kinases and actin. Binding to actin occurs through a C-terminal verprolin homology domain in all family members. The multiprotein complex serves to tranduce signals that involve changes in cell shape, motility or function. The published map location (PMID:10381382) has been changed based on recent genomic sequence comparisons, which indicate that the expressed gene is located on chromosome 1, and a pseudogene may be located on chromosome X. [provided by RefSeq]
Spezifikation
Spezifikation
Zugriffsnummer | NP_008921.1 |
Zur Verwendung mit (Anwendung) | Antibody Production, ELISA, Protein Array, Western Blot |
Zusammensetzung | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gen-ID (Entrez) | 10163 |
Molekulargewicht | 80.7kDa |
Name | WASF2 (Human) Recombinant Protein (P01) |
Reinigungsverfahren | Glutathione Sepharose 4 Fast Flow |
Qualitätskontrollen | 12.5% SDS-PAGE Stained with Coomassie Blue. |
Menge | 25 μg |
Immunogen | MPLVTRNIEPRHLCRQTLPSVRSELECVTNITLANVIRQLGSLSKYAEDIFGELFTQANTFASRVSSLAERVDRLQVKVTQLDPKEEEVSLQGINTRKAFRSSTIQDQKLFDRNSLPVPVLETYNTCDTPPPLNNLTPYRDDGKEALKFYTDPSYFFDLWKEKMLQDTKDIMKEKRKHRKEKKDNPNRGNVNPRKIKTRKEEWEKMKMGQEFVESKEKLGTSGYPPTLVYQNGSIGCVENVDASSYPPPPQSDSASSPSPSFSEDNLPPPPAEFSYPVDNQRGSGLAGPKRSSVVSPSHPPPAPPLGSPPGPKPGFAPPPAPPPPPPPMIGIPPPPPPVGFGSPGTPPPPSPPSFPPHPDFAAPPPPPPPPAADYPTLPPPPLSQPTGGAPPPPPPPPPPGPPPPPFTGADGQPAIPPPLSDTTKPKSSLPAVSDARSDLLSAIRQGFQLRRVEEQREQEKRDVVGNDVATILSRRIAVEYSDSEDDSSEFDEDDWSD |
Mehr anzeigen |
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.