Learn More
Abnova™ Human VPS4A Full-length ORF (NP_037377.1, 1 a.a. - 437 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
361.00 CHF - 547.00 CHF
Spezifikation
Zugriffsnummer | NP_037377.1 |
---|---|
Zur Verwendung mit (Anwendung) | Antibody Production, ELISA, Protein Array, Western Blot |
Zusammensetzung | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gen-ID (Entrez) | 27183 |
Molekulargewicht | 75.3kDa |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
16128652
|
Abnova™
H00027183-P01.25UG |
25 ug |
547.00 CHF
25 Mikrogramm |
Verfügbar ab: 30-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
16118652
|
Abnova™
H00027183-P01.10UG |
10 ug |
361.00 CHF
10 Mikrogramm |
Verfügbar ab: 30-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
Beschreibung
The protein encoded by this gene is a member of the AAA protein family (ATPases associated with diverse cellular activities), and is the homolog of the yeast Vps4 protein. In humans, two paralogs of the yeast protein have been identified. The former share a high degree of aa sequence similarity with each other, and also with yeast Vps4 and mouse Skd1 proteins. The mouse Skd1 (suppressor of K+ transport defect 1) has been shown to be really an yeast Vps4 ortholog. Functional studies indicate that both human paralogs associate with the endosomal compartments, and are involved in intracellular protein trafficking, similar to Vps4 protein in yeast. The gene encoding this paralog has been mapped to chromosome 16; the gene for the other resides on chromosome 18. [provided by RefSeq]
Sequence: MTTSTLQKAIDLVTKATEEDKAKNYEEALRLYQHAVEYFLHAIKYEAHSDKAKESIRAKCVQYLDRAEKLKDYLRSKEKHGKKPVKENQSEGKGSDSDSEGDNPEKKKLQEQLMGAVVMEKPNIRWNDVAGLEGAKEALKEAVILPIKFPHLFTGKRTPWRGILLFGPPGTGKSYLAKAVATEANNSTFFSVSSSDLMSKWLGESEKLVKNLFELARQHKPSIIFIDEVDSLCGSRNENESEAARRIKTEFLVQMQGVGNNNDGTLVLGATNIPWVLDSAIRRRFEKRIYIPLPEEAARAQMFRLHLGSTPHNLTDANIHELARKTEGYSGADISIIVRDSLMQPVRKVQSATHFKKVCGPSRTNPSMMIDDLLTPCSPGDPGAMEMTWMDVPGDKLLEPVVCMSDMLRSLATTRPTVNADDLLKVKKFSEDFGQESSpezifikation
NP_037377.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
75.3kDa | |
Glutathione Sepharose 4 Fast Flow | |
MTTSTLQKAIDLVTKATEEDKAKNYEEALRLYQHAVEYFLHAIKYEAHSDKAKESIRAKCVQYLDRAEKLKDYLRSKEKHGKKPVKENQSEGKGSDSDSEGDNPEKKKLQEQLMGAVVMEKPNIRWNDVAGLEGAKEALKEAVILPIKFPHLFTGKRTPWRGILLFGPPGTGKSYLAKAVATEANNSTFFSVSSSDLMSKWLGESEKLVKNLFELARQHKPSIIFIDEVDSLCGSRNENESEAARRIKTEFLVQMQGVGNNNDGTLVLGATNIPWVLDSAIRRRFEKRIYIPLPEEAARAQMFRLHLGSTPHNLTDANIHELARKTEGYSGADISIIVRDSLMQPVRKVQSATHFKKVCGPSRTNPSMMIDDLLTPCSPGDPGAMEMTWMDVPGDKLLEPVVCMSDMLRSLATTRPTVNADDLLKVKKFSEDFGQES | |
RUO | |
VPS4A | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
27183 | |
VPS4A (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLJ22197/SKD1/SKD2/VPS4/VPS4-1 | |
VPS4A | |
Recombinant | |
wheat germ expression system |