missing translation for 'onlineSavingsMsg'
Learn More

Human VEGFR2/KDR/Flk-1 Alexa Fluor 700-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Artikelnummer. 18733028
Change view
Click to view available options
Menge:
100 μg
Packungsgröße:
100 Mikrogramm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge unitSize
18733028 100 μg 100 Mikrogramm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 18733028 Lieferant R&D Systems Lieferanten-Nr. FAB3572RN100UG

om dit product te kopen Registreer vandaag om een webaccount aan te maken

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Mouse Monoclonal Antibody

VEGFR2/KDR/Flk-1 Monoclonal specifically detects VEGFR2/KDR/Flk-1 in Human samples. It is validated for Flow Cytometry.
TRUSTED_SUSTAINABILITY

Spezifikation

Antigen VEGFR2/KDR/Flk-1
Anwendungen Flow Cytometry
Klassifikation Monoclonal
Klon 89106R
Konjugat Alexa Fluor 700
Verdünnung Flow Cytometry 0.25-1 ug/10^6 cells
Zusammensetzung Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide.
Gen-Zugriffsnummer P35968
Gen-Alias CD309, CD309 antigen, EC 2.7.10, EC 2.7.10.1, Fetal liver kinase 1, fetal liver kinase-1, Flk1, Flk-1, FLK1tyrosine kinase growth factor receptor, KDR, Kinase insert domain receptor, kinase insert domain receptor (a type III receptor tyrosine kinase), KRD1, Ly73, Protein-tyrosine kinase receptor flk-1, soluble VEGFR2, vascular endothelial growth factor receptor 2, VEGF R2, VEGFR, VEGFR2, VEGFR-2
Wirtsspezies Mouse
Immunogen S. frugiperda insect ovarian cell line Sf 21-derived recombinant human VEGFR2/KDR/Flk-1, Ala20-Glu764, Accession # P35968
Reinigungsverfahren Protein A or G purified from cell culture supernatant
Menge 100 μg
Regulatorischer Status RUO
Primär oder sekundär Primary
Gen-ID (Entrez) 3791
Zielspezies Human
Inhalt und Lagerung Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied.
Form Purified
Isotype IgG1
Mehr anzeigen Weniger anzeigen
Name des Produkts
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.