missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human VEGFR2/KDR/Flk-1 Alexa Fluor 405-conjugated Antibody, R&D Systems™

Beschreibung
VEGFR2/KDR/Flk-1 Monoclonal specifically detects VEGFR2/KDR/Flk-1 in Human samples. It is validated for Flow Cytometry.
Spezifikation
Spezifikation
| Antigen | VEGFR2/KDR/Flk-1 |
| Anwendungen | Flow Cytometry |
| Klassifikation | Monoclonal |
| Klon | 89106R |
| Konjugat | Alexa Fluor 405 |
| Verdünnung | Flow Cytometry 0.25-1 ug/10^6 cells |
| Zusammensetzung | Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. |
| Gen-Zugriffsnummer | P35968 |
| Gen-Alias | CD309, CD309 antigen, EC 2.7.10, EC 2.7.10.1, Fetal liver kinase 1, fetal liver kinase-1, Flk1, Flk-1, FLK1tyrosine kinase growth factor receptor, KDR, Kinase insert domain receptor, kinase insert domain receptor (a type III receptor tyrosine kinase), KRD1, Ly73, Protein-tyrosine kinase receptor flk-1, soluble VEGFR2, vascular endothelial growth factor receptor 2, VEGF R2, VEGFR, VEGFR2, VEGFR-2 |
| Wirtsspezies | Mouse |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?