Learn More
ACROBiosystems Human VEGF-B Protein, His Tag
Marke: ACROBiosystems VE6-H5225-50UG
Additional Details : Gewicht : 0.00001kg
Beschreibung
Vascular endothelial growth factor B (VEGFB) is also known as VEGF-related factor (VRF), is a secreted O-glycosylated protein, which belongs to the PDGF/VEGF growth factor family. VEGFB is expressed in all tissues except liver. Highest levels found in heart, skeletal muscle and pancreas. VEGFB is growth factor for endothelial cells. VEGF-B seems to play a role only in the maintenance of newly formed blood vessels during pathological conditions. VEGF-B also plays an important role on several types of neurons. It is important for the protection of neurons in the retina and the cerebral cortex during stroke and of motoneurons during motor neuron diseases such as amyotrophic lateral sclerosis.VEGF-B167 binds heparin and neuropilin-1 whereas the binding to neuropilin-1 of VEGF-B186 is regulated by proteolysis.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 20.3 kDa. The protein migrates as 15 kDa and 30-35 kDa under reducing (R) condition (SDS-PAGE).Spezifikation
NP_003368.1 | |
50 µg | |
Pro 22 - Ala 207 | |
VRF,VEGFL,VEGFB,VEGF-B | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / µg by the LAL method. | |
Powder | |
>90% as determined by SDS-PAGE |
20.3 kDa | |
Human VEGF-B, His Tag (VE6-H5225) is expressed from human 293 cells (HEK293). It contains AA Pro 22 - Ala 207 (Accession # P49765-1). | |
-20°C | |
VEGF-B | |
Human | |
PBS, pH7.4 | |
HEK293 | |
Cytokines and Growth Factors |