Learn More
Abnova™ Human UPF3A Partial ORF (NP_075387, 382 a.a. - 475 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marke: Abnova™ H00065110-Q01.10ug
Additional Details : Gewicht : 0.00010kg
Beschreibung
This gene encodes a protein that is part of a post-splicing multiprotein complex involved in both mRNA nuclear export and mRNA surveillance. The encoded protein is one of two functional homologs to yeast Upf3p. mRNA surveillance detects exported mRNAs with truncated open reading frames and initiates nonsense-mediated mRNA decay (NMD). When translation ends upstream from the last exon-exon junction, this triggers NMD to degrade mRNAs containing premature stop codons. This protein binds to the mRNA and remains bound after nuclear export, acting as a nucleocytoplasmic shuttling protein. It forms with Y14 a complex that binds specifically 20 nt upstream of exon-exon junctions. This gene is located on the long arm of chromosome 13. Two splice variants encoding different isoforms have been found for this gene. [provided by RefSeq]
Sequence: SEDEQRWGKGPGQDRGKKGSQDSGAPGEAMERLGRAQRCDDSPAPRKERLANKDRPALQLYDPGARFRARECGGNRRICKAEGSGTGPEKREEASpezifikation
NP_075387 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.08kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
SEDEQRWGKGPGQDRGKKGSQDSGAPGEAMERLGRAQRCDDSPAPRKERLANKDRPALQLYDPGARFRARECGGNRRICKAEGSGTGPEKREEA | |
RUO | |
UPF3A | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
65110 | |
UPF3A (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
HUPF3A/RENT3A/UPF3 | |
UPF3A | |
Recombinant | |
wheat germ expression system |