Learn More
Abnova™ Human UNC5C Partial ORF (AAH41156, 498 a.a. - 597 a.a.) Recombinant Protein with GST-tag at N-terminal
Beschreibung
This gene product belongs to the UNC-5 family of netrin receptors. Netrins are secreted proteins that direct axon extension and cell migration during neural development. They are bifunctional proteins that act as attractants for some cell types and as repellents for others, and these opposite actions are thought to be mediated by two classes of receptors. The UNC-5 family of receptors mediate the repellent response to netrin; they are transmembrane proteins containing 2 immunoglobulin (Ig)-like domains and 2 type I thrombospondin motifs in the extracellular region. [provided by RefSeq]
Spezifikation
Spezifikation
Zugriffsnummer | AAH41156 |
Zur Verwendung mit (Anwendung) | Antibody Production, ELISA, Protein Array, Western Blot |
Zusammensetzung | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gen-ID (Entrez) | 8633 |
Molekulargewicht | 36.41kDa |
Name | UNC5C (Human) Recombinant Protein (Q01) |
Qualitätskontrollen | 12.5% SDS-PAGE Stained with Coomassie Blue. |
Menge | 25 μg |
Immunogen | IKVYNTSGAVTPQDDLSEFTSKLSPQMTQSLLENEALSLKNQSLARQTDPSCTAFGSFNSLGGHLIVPNSGVSLLIPAGAIPQGRVYEMYVTVHRKETMR |
Lagerungsbedingungen | Store at -80°C. Aliquot to avoid repeated freezing and thawing. |
Mehr anzeigen |
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.