missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human ULBP-1 DuoSet ELISA
Description
The Human ULBP-1 DuoSet ELISA is a Solid Phase Sandwich ELISA that quantifies human ULBP-1.
Specifications
Specifications
| Produkttyp | Solid Phase Sandwich ELISA Development Kit |
| Menge | 1 Kit (for 15 Plates) |
| Spezies | Human |
| Konjugat | Biotin |
| Assay-Bereich | 93.80 to 6000 pg/mL |
| Zur Verwendung mit (Anwendung) | ELISA |
| Gen-ID (Entrez) | 80329 |
| Produktlinie | R&D Systems |
| Inhalt und Lagerung | Store the unopened product at 2°C to 8°C. Do not use past expiration date. |
| Research Area | Immunology |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?