missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human UBXD5 Partial ORF (NP_663320.1, 311 a.a. - 409 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marke:  Abnova™ H00091544-Q01.25ug

Additional Details : Gewicht : 0.00010kg

 Lernen Sie weitere Varianten dieses

Artikelnummer. 16131167

  • 547.00 CHF / 25 Mikrogramm
Verfügbar ab: 25-06-2024
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

This gene encodes a protein with a divergent C-terminal UBX domain. The homologous protein in the rat interacts with members of the Rnd subfamily of Rho GTPases at the cell periphery through its C-terminal region. It also interacts with several heterotrimeric G proteins through their G-alpha subunits and promotes Rho GTPase activation. It is proposed to serve a bidirectional role in the promotion and inhibition of Rho activity through upstream signaling pathways. The 3' coding sequence of this gene contains a polymoprhic region of 24 nt tandem repeats. Several transcripts containing between 1.5 and five repeat units have been reported. Multiple transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]

Sequence: QGEVIDIRGPIRDTLQNCCPLPARIQEIVVETPTLAAERERSQESPNTPAPPLSMLRIKSENGEQAFLLMMQPDNTIGDVRALLAQARVMDASAFEIFS
Spezifikation

Spezifikation

NP_663320.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.41kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
QGEVIDIRGPIRDTLQNCCPLPARIQEIVVETPTLAAERERSQESPNTPAPPLSMLRIKSENGEQAFLLMMQPDNTIGDVRALLAQARVMDASAFEIFS
RUO
UBXN11
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
91544
UBXD5 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
COA-1/DKFZp686F04228/PP2243/SOC/SOCI/UBXD5
UBXN11
Recombinant
wheat germ expression system
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote