Learn More
ACROBiosystems Human TrkB / NTRK2 Protein, His Tag
Marke: ACROBiosystems NT2-H5228-100UG
Additional Details : Gewicht : 0.00001kg
Beschreibung
Neurotrophic tyrosine kinase receptor type 2 (NTRK2) is also known as BDNF/NT-3 growth factors receptor, Tropomyosin-related kinase B (TRKB) and TrkB tyrosine kinase, which belongs to the protein kinase superfamily or Tyr protein kinase family. Insulin receptor subfamily. NTRK2 / TrkB contains two Ig-like C2-type (immunoglobulin-like) domains, two LRR (leucine-rich) repeats, one LRRCT domain, one LRRNT domain, one protein kinase domain. NTRK2 / Trk-B is expressed in the central and peripheral nervous system. The catalytic activity of NTRK2 is “ATP + a [protein]-L-tyrosine = ADP + a [protein]-L-tyrosine phosphate”. NTRK2 / TrkB involved in the development and the maturation of the central and the peripheral nervous systems through regulation of neuron survival, proliferation, migration, differentiation, and synapse formation and plasticity.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 45.2 kDa. The protein migrates as 60-90 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spezifikation
AAH31835 | |
100 μg | |
Cys 32 - His 430 | |
NTRK2,TRKB,GP145-TrkB | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |
45.2 kDa | |
Human TrkB, His Tag (NT2-H5228) is expressed from human 293 cells (HEK293). It contains AA Cys 32 - His 430 (Accession # AAH31835). | |
-20°C | |
TrkB | |
Human | |
PBS, pH7.4 | |
HEK293 | |
Bio-Markers and CD Antigens |