missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human TRIM35 Full-length ORF (NP_741983.2, 1 a.a. - 493 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marke:  Abnova™ H00023087-P01.10ug

Additional Details : Gewicht : 0.00010kg

 Lernen Sie weitere Varianten dieses

Artikelnummer. 16126932

  • 379.00 CHF / 10 Mikrogramm
Verfügbar ab: 09-01-2024
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

The protein encoded by this gene is a member of the tripartite motif (TRIM) family. The TRIM motif includes three zinc-binding domains, a RING, a B-box type 1 and a B-box type 2, and a coiled-coil region. The function of this protein has not been identified. [provided by RefSeq]

Sequence: MERSPDVSPGPSRSFKEELLCAVCYDPFRDAVTLRCGHNFCRGCVSRCWEVQVSPTCPVCKDRASPADLRTNHTLNNLVEKLLREEAEGARWTSYRFSRVCRLHRGQLSLFCLEDKELLCCSCQADPRHQGHRVQPVKDTAHDFRAKCRNMEHALREKAKAFWAMRRSYEAIAKHNQVEAAWLEGRIRQEFDKLREFLRVEEQAILDAMAEETRQKQLLADEKMKQLTEETEVLAHEIERLQMEMKEDDVSFLMKHKSRKRRLFCTMEPEPVQPGMLIDVCKYLGSLQYRVWKKMLASVESVPFSFDPNTAAGWLSVSDDLTSVTNHGYRVQVENPERFSSAPCLLGSRVFSQGSHAWEVALGGLQSWRVGVVRVRQDSGAEGHSHSCYHDTRSGFWYVCRTQGVEGDHCVTSDPATSPLVLAIPRRLRVELECEEGELSFYDAERHCHLYTFHARFGEVRPYFYLGGARGAGPPEPLRICPLHISVKEELDG
Spezifikation

Spezifikation

NP_741983.2
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
82.9kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
HLS5/KIAA1098/MAIR/MGC17233
TRIM35
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
23087
TRIM35 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MERSPDVSPGPSRSFKEELLCAVCYDPFRDAVTLRCGHNFCRGCVSRCWEVQVSPTCPVCKDRASPADLRTNHTLNNLVEKLLREEAEGARWTSYRFSRVCRLHRGQLSLFCLEDKELLCCSCQADPRHQGHRVQPVKDTAHDFRAKCRNMEHALREKAKAFWAMRRSYEAIAKHNQVEAAWLEGRIRQEFDKLREFLRVEEQAILDAMAEETRQKQLLADEKMKQLTEETEVLAHEIERLQMEMKEDDVSFLMKHKSRKRRLFCTMEPEPVQPGMLIDVCKYLGSLQYRVWKKMLASVESVPFSFDPNTAAGWLSVSDDLTSVTNHGYRVQVENPERFSSAPCLLGSRVFSQGSHAWEVALGGLQSWRVGVVRVRQDSGAEGHSHSCYHDTRSGFWYVCRTQGVEGDHCVTSDPATSPLVLAIPRRLRVELECEEGELSFYDAERHCHLYTFHARFGEVRPYFYLGGARGAGPPEPLRICPLHISVKEELDG
RUO
TRIM35
Wheat Germ (in vitro)
GST
Liquid
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote