missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human TRAIL R2/TNFRSF10B Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 26 publications
Marke: R&D Systems AF631
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
TRAILR2/TNFRSF10B Polyclonal specifically detects TRAILR2/TNFRSF10B in Human samples. It is validated for Western Blot, Agonist Activity.
Spezifikation
| TRAILR2/TNFRSF10B | |
| Polyclonal | |
| Unconjugated | |
| Q6FH58 | |
| TNFRSF10B | |
| Mouse myeloma cell line NS0-derived recombinant human TRAIL R2/TNFRSF10B Ala54-Glu182 Accession # Q6FH58 | |
| 100 μg | |
| Agonist Activity | |
| 8795 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot | |
| LYOPH | |
| Western Blot 1 ug/mL, Agonist Activity | |
| CD262, CD262 antigen, death domain containing receptor for TRAIL/Apo-2L, Death receptor 5, DR5, DR5TRICK2B, Fas-like protein, KILLER/DR5, KILLERapoptosis inducing protein TRICK2A/2B, TNF-related apoptosis-inducing ligand receptor 2, TNFRSF10B, TRAIL receptor 2, TRAILR2, TRAIL-R2apoptosis inducing receptor TRAIL-R2, TRAILR2cytotoxic TRAIL receptor-2, TRICK2, TRICK2A, TRICKB, tumor necrosis factor receptor superfamily member 10B, tumor necrosis factor receptor superfamily, member 10b, tumor necrosis factor receptor-like protein ZTNFR9, ZTNFR9 | |
| Goat | |
| Antigen Affinity-purified | |
| RUO | |
| Primary | |
| Detects human TRAIL R2/TNFRSF10B in direct ELISAs and Western blots. | |
| Human | |
| IgG |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur