missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Human TPO (Thrombopoietin) Recombinant Protein, PeproTech®
Click to view available options
Menge:
1 mg
10 μg
100 μg
2 μg
250 μg
50 μg
500 μg
Packungsgröße:
1 Milligramm
10 Mikrogramm
100 Mikrogramm
2 Mikrogramm
250 Mikrogramm
50 Mikrogramm
500 Mikrogramm
Beschreibung
Recombinant Human TPO is a fully biologically active 174 amino acid polypeptide (18.6 kDa), which contains the erythropoietin-like domain of the full length TPO protein. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis
Thrombopoietin (TPO, THPO, MPL ligand, megakaryocyte colony-stimulating factor) is responsible for the generation of platelets through its action on hematopoietic cells committed to the megakaryocyte lineage. Binding of thrombopoietin to its receptor, MPL (CD110), initiates a signaling cascade via JAK/STAT, MAPK, and PI3K/AKT signaling pathways that leads to the proliferation and differentiation of megakaryocytes and, ultimately, to the production of platelets (thrombopoiesis). Thrombopoietin is constitutively produced by the liver and has a predicted molecular weight of 38 kDa.
Spezifikation
Spezifikation
| Zugriffsnummer | P40225 |
| Zur Verwendung mit (Anwendung) | ELISA, Functional Assay, Western Blot |
| Zusammensetzung | protein with no preservative |
| Gen-ID (Entrez) | 7066 |
| Molekulargewicht | 18.6 kDa |
| Name | Human TPO (Thrombopoietin) |
| Menge | 250 μg |
| Quelle | E. coli |
| Kennzeichnung | RUO |
| Endotoxin-Konzentration | <1 EU/ μg |
| Mehr anzeigen |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur