missing translation for 'onlineSavingsMsg'
Learn More

ACROBiosystems Human TNFR1 / CD120a / TNFRSF1A Protein, Fc Tag

Marke:  ACROBiosystems TN1-H5251-1MG

Additional Details : Gewicht : 0.00001kg

 Lernen Sie weitere Varianten dieses

Artikelnummer. 15827807

  • 1842.35 CHF / 1 Milligramm


Verfügbar ab
28-05-2021
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

Tumor necrosis factor receptor 1 (TNF-R1) is also known as Tumor necrosis factor receptor superfamily member 1A (TNFRSF1A), TNFAR, CD antigen CD120a, which belongs to the tumor necrosis factor receptor superfamily. TNF-R1 contains one death domain and four TNFR-Cys repeats. TNF-R1 is the receptor of TNFSF2 / TNF-alpha and homotrimeric TNFSF1 / lymphotoxin - alpha. The adapter molecule FADD recruits caspase-8 to the activated receptor. The resulting death-inducing signaling complex (DISC) performs caspase-8 proteolytic activation which initiates the subsequent cascade of caspases (aspartate-specific cysteine proteases) mediating apoptosis. TNF-R1 contributes to the induction of non - cytocidal TNF effects including anti-viral state and activation of the acid sphingomyelinase. Defects in TNFRSF1A are the cause of familial hibernian fever (FHF).

This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 47.9 kDa. The protein migrates as 58-66 kDa under reducing (R) condition (SDS-PAGE) due to different glycosylation.
Spezifikation

Spezifikation

NP_001056
HEK293
Human
47.9 kDa
1 mg
Ile 22 - Thr 211
TNFR1,CD120a,TNFRSF1A,FPF,TBP1,TNF-R,TNF-R-I,TNF-R55,TNFAR,TNFR55,TNFR60,p55,p60
Unconjugated
Tris with Glycine, Arginine and NaCl, pH7.5
Less than 1.0 EU / µg by the LAL method.
Bio-Markers and CD Antigens
Powder
>98% as determined by SDS-PAGE
Human TNFR1, Fc Tag (TN1-H5251) is expressed from human 293 cells (HEK293). It contains AA Ile 22 - Thr 211 (Accession # NP_001056).
-20°C
TNFR1
Fc Tag
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation