missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human TNFAIP3 Full-length ORF (AAH98379.1, 1 a.a. - 248 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
auf Anfrage
Spezifikation
Name | TNFAIP3 (Human) Recombinant Protein (P01) |
---|---|
Rekombinant | Yes |
Reinigungsverfahren | Glutathione Sepharose 4 Fast Flow |
Molekulargewicht | 55.3kDa |
Lagerungsbedingungen | Store at -80°C. Aliquot to avoid repeated freezing and thawing. |
Beschreibung
This gene was identified as a gene whose expression is rapidly induced by the tumor necrosis factor (TNF). The protein encoded by this gene is a zinc finger protein, and has been shown to inhibit NF-kappa B activation as well as TNF-mediated apoptosis. Knockout studies of a similar gene in mice suggested that this gene is critical for limiting inflammation by terminating TNF-induced NF-kappa B responses. [provided by RefSeq]
Sequence: MAEQVLPQALYLSNMRKAVKIRERTPEDIFKPTNGIIHHFKTMHRYTLEMFRTCQFCPQFREIIHKALIDRNIQATLESQKKLNWCREVRKLVALKTNGDGNCLMHATSQYMWGVQDTDLVLRKALFSTLKETDTRNFKFRWQLESLKSQEFVETGLCYDTRNWNDEWDNLIKMASTDTPMARSGLQYNSLEEIHIFVLCNILRRPIIVISGEMPADHGSVLKCFQPYALAPGENHTAKVQVTEFNGISpezifikation
TNFAIP3 (Human) Recombinant Protein (P01) | |
Glutathione Sepharose 4 Fast Flow | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. |
Yes | |
55.3kDa |