missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human TNFAIP3 Full-length ORF (AAH98379.1, 1 a.a. - 248 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marke:  Abnova™ H00007128-P01.25ug

Additional Details : Gewicht : 0.00010kg

 Lernen Sie weitere Varianten dieses

Artikelnummer. 16151472

  • 448.00 CHF / 25 Mikrogramm


Verfügbar ab
07-01-2020
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

Sequence: MAEQVLPQALYLSNMRKAVKIRERTPEDIFKPTNGIIHHFKTMHRYTLEMFRTCQFCPQFREIIHKALIDRNIQATLESQKKLNWCREVRKLVALKTNGDGNCLMHATSQYMWGVQDTDLVLRKALFSTLKETDTRNFKFRWQLESLKSQEFVETGLCYDTRNWNDEWDNLIKMASTDTPMARSGLQYNSLEEIHIFVLCNILRRPIIVISGEMPADHGSVLKCFQPYALAPGENHTAKVQVTEFNGI
Spezifikation

Spezifikation

AAH98379.1
Liquid
7128
TNFAIP3 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MAEQVLPQALYLSNMRKAVKIRERTPEDIFKPTNGIIHHFKTMHRYTLEMFRTCQFCPQFREIIHKALIDRNIQATLESQKKLNWCREVRKLVALKTNGDGNCLMHATSQYMWGVQDTDLVLRKALFSTLKETDTRNFKFRWQLESLKSQEFVETGLCYDTRNWNDEWDNLIKMASTDTPMARSGLQYNSLEEIHIFVLCNILRRPIIVISGEMPADHGSVLKCFQPYALAPGENHTAKVQVTEFNGI
RUO
TNFAIP3
wheat germ expression system
Yes
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
55.3kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
A20|MGC104522|MGC138687|MGC138688|OTUD7C|TNFA1P2
TNFAIP3
Wheat Germ (in vitro)
GST
Videos
Dokumentation