Learn More
ACROBiosystems Human TNF-alpha Protein, His Tag (active trimer)
MALS verified
Marke: ACROBiosystems TNA-H5228-100UG
Additional Details : Gewicht : 0.00001kg
Beschreibung
Tumor necrosis factor alpha (TNFα) is a cytokine produced primarily by monocytes and macrophages. It is found in synovial cells and macrophages in the tissues.The primary role of TNFα is in the regulation of immune cells. TNFα is able to induce apoptotic cell death, to induce inflammation, and to inhibit tumorigenesis and viral replication. Dysregulation of TNFα production has been implicated in a variety of human diseases, including major depression, Alzheimer's disease and cancer. Recombinant TNFα is used as an immunostimulant under the INN tasonermin. TNFα can be produced ectopically in the setting of malignancy and parallels parathyroid hormone both in causing secondary hypercalcemia and in the cancers with which excessive production is associated.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 18.3 kDa. The protein migrates as 18.2 kDa under reducing (R) condition (SDS-PAGE).Spezifikation
NP_000585.2 | |
100 μg | |
Val 77 - Leu 233 | |
DIF,TNF-alpha,TNFA,TNFSF2,cachexin,cachectin,TNFα | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |
18.3 kDa | |
Human TNF-alpha, His Tag (active trimer) (MALS verified) (TNA-H5228) is expressed from human 293 cells (HEK293). It contains AA Val 77 - Leu 233 (Accession # NP_000585.2). | |
-20°C | |
TNF-alpha | |
Human | |
PBS, pH7.4 | |
HEK293 | |
Cytokines and Growth Factors |