missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human TMEM176B Full-length ORF (NP_054739.2, 1 a.a. - 270 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marke: Abnova™ H00028959-P01.10ug
Additional Details : Gewicht : 0.00010kg
Beschreibung
Sequence: MTQNTVIVNGVAMASRPSQPTHVNVHIHQESALTQLLKAGGSLKKFLFHPGDTVPSTARIGYEQLALGVTQILLGVVSCVLGVCLSLGPWTVLRASGCAFWAGSVVIAAGAGAIVHEKHPGKLAGYISSLLTLTGFATAMAAVVLCVNSFIWQTEPFLYIDTVCDRSDPVFPTTGYRWMRRSQENQWQKEECRAYMQMLRKLFTAIRALFLAVCVLKVIVSLVSLGVGLRNLCGQSSQPLNEEGSEKRLLGENSVPPSPSREQTSTAIVLSpezifikation
NP_054739.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
55.6kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLJ26014/LR8/MGC110857 | |
TMEM176B | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
28959 | |
TMEM176B (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MTQNTVIVNGVAMASRPSQPTHVNVHIHQESALTQLLKAGGSLKKFLFHPGDTVPSTARIGYEQLALGVTQILLGVVSCVLGVCLSLGPWTVLRASGCAFWAGSVVIAAGAGAIVHEKHPGKLAGYISSLLTLTGFATAMAAVVLCVNSFIWQTEPFLYIDTVCDRSDPVFPTTGYRWMRRSQENQWQKEECRAYMQMLRKLFTAIRALFLAVCVLKVIVSLVSLGVGLRNLCGQSSQPLNEEGSEKRLLGENSVPPSPSREQTSTAIVL | |
RUO | |
TMEM176B | |
Wheat Germ (in vitro) | |
GST | |
Liquid |