missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human TLL2 Full-length ORF (AAI11600.1, 1 a.a. - 1015 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
361.00 CHF - 547.00 CHF
Spezifikation
Zugriffsnummer | AAI11600.1 |
---|---|
Zur Verwendung mit (Anwendung) | Antibody Production, Protein Array, ELISA, Western Blot |
Zusammensetzung | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gen-ID (Entrez) | 7093 |
Molekulargewicht | 138.6kDa |
Product Code | Brand | Menge | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Menge | Price | Quantity & Availability | |||||
16161432
|
Abnova™
H00007093-P01.25UG |
25 ug |
547.00 CHF
25 Mikrogramm |
Verfügbar ab: 30-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
16151432
|
Abnova™
H00007093-P01.10UG |
10 ug |
361.00 CHF
10 Mikrogramm |
Verfügbar ab: 30-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
Description
This gene encodes an astacin-like zinc-dependent metalloprotease and is a subfamily member of the metzincin family. Unlike other family members, a similar protein in mice does not cleave procollagen C-propeptides or chordin. [provided by RefSeq]
Sequence: MPRATALGALVSLLLLLPLPRGAGGLGERPDATADYSELDGEEGTEQQLEHYHDPCKAAVFWGDIALDEDDLKLFHIDKARDWTKQTVGATGHSTGGLEEQASESSPDTTAMDTGTKEAGKDGRENTTLLHSPGTLHAAAKTFSPRVRRATTSRTERIWPGGVIPYVIGGNFTGSQRAIFKQAMRHWEKHTCVTFIERTDEESFIVFSYRTCGCCSYVGRRGGGPQAISIGKNCDKFGIVAHELGHVVGFWHEHTRPDRDQHVTIIRENIQPGQEYNFLKMEAGEVSSLGETYDFDSIMHYARNTFSRGVFLDTILPRQDDNGVRPTIGQRVRLSQGDIAQARKLYKCPACGETLQDTTGNFSAPGFPNGYPSYSHCVWRISVTPGEKIVLNFTSMDLFKSRLCWYDYVEVRDGYWRKAPLLGRFCGDKIPEPLVSTDSRLWVEFRSSSNILGKGFFAAYEATCGGDMNKDAGQIQSPNYPDDYRPSKECVWRITVSEGFHVGLTFQAFEIERHDSCAYDYLEVRDGPTEESALIGHFCGYEKPEDVKSSSNRLWMKFVSDGSINKAGFAANFFKEVDECSWPDHGGCEHRCVNTLGSYKCACDPGYELAADKKMCEVACGGFITKLNGTITSPGWPKEYPTNKNCVWQVVAPAQYRISLQFEVFELEGNDVCKYDFVEVRSGLSPDAKLHGRFCGSETPEVITSQSNNMRVEFKSDNTVSKRGFRAHFFSDKDECAKDNGGCQHECVNTFGSYLCRCRNGYWLHENGHDCKEAGCAHKISSVEGTLASPNWPDKYPSRRECTWNISSTAGHRVKLTFNEFEIEQHQECAYDHLEMYDGPDSLAPILGRFCGSKKPDPTVASGSSMFLRFYSDASVQRKGFQAVHSTECGGRLKAEVQTKELYSHAQFGDNNYPSEARCDWVIVAEDGYGVELTFRTFEVEEEADCGYDYMEAYDGYDSSAPRLGRFCGSGPLEEIYSAGDSLMIRFRTDDTINKKGFHARYTSTKFQDALHMKKSpecifications
AAI11600.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
138.6kDa | |
Glutathione Sepharose 4 Fast Flow | |
MPRATALGALVSLLLLLPLPRGAGGLGERPDATADYSELDGEEGTEQQLEHYHDPCKAAVFWGDIALDEDDLKLFHIDKARDWTKQTVGATGHSTGGLEEQASESSPDTTAMDTGTKEAGKDGRENTTLLHSPGTLHAAAKTFSPRVRRATTSRTERIWPGGVIPYVIGGNFTGSQRAIFKQAMRHWEKHTCVTFIERTDEESFIVFSYRTCGCCSYVGRRGGGPQAISIGKNCDKFGIVAHELGHVVGFWHEHTRPDRDQHVTIIRENIQPGQEYNFLKMEAGEVSSLGETYDFDSIMHYARNTFSRGVFLDTILPRQDDNGVRPTIGQRVRLSQGDIAQARKLYKCPACGETLQDTTGNFSAPGFPNGYPSYSHCVWRISVTPGEKIVLNFTSMDLFKSRLCWYDYVEVRDGYWRKAPLLGRFCGDKIPEPLVSTDSRLWVEFRSSSNILGKGFFAAYEATCGGDMNKDAGQIQSPNYPDDYRPSKECVWRITVSEGFHVGLTFQAFEIERHDSCAYDYLEVRDGPTEESALIGHFCGYEKPEDVKSSSNRLWMKFVSDGSINKAGFAANFFKEVDECSWPDHGGCEHRCVNTLGSYKCACDPGYELAADKKMCEVACGGFITKLNGTITSPGWPKEYPTNKNCVWQVVAPAQYRISLQFEVFELEGNDVCKYDFVEVRSGLSPDAKLHGRFCGSETPEVITSQSNNMRVEFKSDNTVSKRGFRAHFFSDKDECAKDNGGCQHECVNTFGSYLCRCRNGYWLHENGHDCKEAGCAHKISSVEGTLASPNWPDKYPSRRECTWNISSTAGHRVKLTFNEFEIEQHQECAYDHLEMYDGPDSLAPILGRFCGSKKPDPTVASGSSMFLRFYSDASVQRKGFQAVHSTECGGRLKAEVQTKELYSHAQFGDNNYPSEARCDWVIVAEDGYGVELTFRTFEVEEEADCGYDYMEAYDGYDSSAPRLGRFCGSGPLEEIYSAGDSLMIRFRTDDTINKKGFHA | |
RUO | |
TLL2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
7093 | |
TLL2 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
KIAA0932/MGC133312/MGC133313 | |
TLL2 | |
Recombinant | |
wheat germ expression system |