missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human TCP-1 beta (aa 226-365) Control Fragment Recombinant Protein

Artikelnummer. 30201457
Ansicht ändern
Click to view available options
Menge:
100 μl
Packungsgröße:
100 Mikroliter
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge Packungsgröße
30201457 100 μl 100 Mikroliter
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 30201457 Lieferant Invitrogen™ Lieferanten-Nr. RP96784
 Eingeschränkter Artikel
Verfügbar ab: 16-12-2026
Zum Warenkorb hinzufügen
Zum Warenkorb hinzufügen

missing translation for 'validMainframeMsg'

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (100%), Rat (100%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-82069. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

TCP-1 beta is a molecular chaperone that is member of the chaperonin containing TCP1 complex (CCT), also known as the TCP1 ring complex (TRiC). The CCT complex consists of two identical stacked rings, each containing eight different proteins. Unfolded polypeptides enter the central cavity of the complex and are folded in an ATP-dependent manner. The complex folds various proteins, including actin and tubulin. Alternate transcriptional splice variants of the TCP-1 beta gene have been observed but not thoroughly characterized. A disease associated with TCP-1 beta protein dysfunction include epididymo-orchitis.
TRUSTED_SUSTAINABILITY

Spezifikation

Zugriffsnummer P78371
Konzentration ≥5.0 mg/mL
Zur Verwendung mit (Anwendung) Neutralization, Control
Zusammensetzung PBS, 1M urea with no preservative; pH 7.4
Gen-ID (Entrez) 10576
Name Human TCP-1 beta (aa 226-365) Control Fragment
pH-Bereich 7.4
Reinigungsverfahren Purified
Menge 100 μl
Lagerungsbedingungen -20°C, Avoid Freeze/Thaw Cycles
Regulatorischer Status RUO
Gen-Alias 99D8.1; AI528772; c-cpn; CCT; Cct1; CCT2; Ccta; CCTB; CCT-beta; chaperonin containing t-complex polypeptide 1, beta subunit; chaperonin containing t-complex polypeptide 1, subunit 2; chaperonin containing TCP1 subunit 2; chaperonin containing Tcp1, subunit 2 (beta); chaperonin subunit 2 (beta); chaperonin-containing T-complex polypeptide beta subunit; epididymis secretory sperm binding protein Li 100n; HEL-S-100n; hypothetical protein LOC505313; p63; PRO1633; T-complex protein 1 subunit beta; T-complex protein 1, beta subunit; Tcp-1; TCP-1-beta; Tp63; TRic
Gebräuchliche Bezeichnung TCP-1 beta
Gensymbol CCT2
Produkttyp Protein
Konjugat Unconjugated
Spezies Human
Rekombinant Recombinant
Proteinmarkierung His-ABP-tag
Sequenz VNQPKRIENAKILIANTGMDTDKIKIFGSRVRVDSTAKVAEIEHAEKEKMKEKVERILKHGINCFINRQLIYNYPEQLFGAAGVMAIEHADFAGVERLALVTGGEIASTFDHPELVKLGSCKLIEEVMIGEDKLIHFSGV
Inhalt und Lagerung -20°C, Avoid Freeze/Thaw Cycles
Expressionssystem E. coli
Form Liquid
Reinheits- oder Qualitätsgrad >80% by SDS-PAGE and Coomassie blue staining
Mehr anzeigen Weniger anzeigen
Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.