Learn More
Abnova™ Human TCEB3 Partial ORF (NP_003189, 81 a.a. - 190 a.a.) Recombinant Protein with GST-tag at N-terminal
Beschreibung
This gene encodes the protein elongin A, which is a subunit of the transcription factor B (SIII) complex. The SIII complex is composed of elongins A/A2, B and C. It activates elongation by RNA polymerase II by suppressing transient pausing of the polymerase at many sites within transcription units. Elongin A functions as the transcriptionally active component of the SIII complex, whereas elongins B and C are regulatory subunits. Elongin A2 is specifically expressed in the testis, and capable of forming a stable complex with elongins B and C. The von Hippel-Lindau tumor suppressor protein binds to elongins B and C, and thereby inhibits transcription elongation. [provided by RefSeq]
Spezifikation
Spezifikation
Zugriffsnummer | NP_003189 |
Zur Verwendung mit (Anwendung) | Antibody Production, ELISA, Protein Array, Western Blot |
Zusammensetzung | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gen-ID (Entrez) | 6924 |
Molekulargewicht | 37.84kDa |
Name | TCEB3 (Human) Recombinant Protein (Q01) |
Qualitätskontrollen | 12.5% SDS-PAGE Stained with Coomassie Blue. |
Menge | 10 µg |
Immunogen | NAEPDEQDFEKSNSRKRPRDALQKEEEMEGDYQETWKATGSRSYSPDHRQKKHRKLSELERPHKVSHGHERRDERKRCHRMSPTYSSDPESSDYGHVQSPPSCTSPHQMY |
Lagerungsbedingungen | Store at -80°C. Aliquot to avoid repeated freezing and thawing. |
Mehr anzeigen |
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.