Learn More
Abnova™ Human TBL1X Full-length ORF (AAH32708.1, 1 a.a. - 526 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
361.00 CHF - 547.00 CHF
Spezifikation
Zugriffsnummer | AAH32708.1 |
---|---|
Zur Verwendung mit (Anwendung) | Antibody Production, Protein Array, ELISA, Western Blot |
Zusammensetzung | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gen-ID (Entrez) | 6907 |
Molekulargewicht | 83.4kDa |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
16131232
|
Abnova™
H00006907-P01.25UG |
25 ug |
547.00 CHF
25 Mikrogramm |
Verfügbar ab: 30-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
16121232
|
Abnova™
H00006907-P01.10UG |
10 ug |
361.00 CHF
10 Mikrogramm |
Verfügbar ab: 30-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
Beschreibung
The protein encoded by this gene has sequence similarity with members of the WD40 repeat-containing protein family. The WD40 group is a large family of proteins, which appear to have a regulatory function. It is believed that the WD40 repeats mediate protein-protein interactions and members of the family are involved in signal transduction, RNA processing, gene regulation, vesicular trafficking, cytoskeletal assembly and may play a role in the control of cytotypic differentiation. This encoded protein is found as a subunit in corepressor SMRT (silencing mediator for retinoid and thyroid receptors) complex along with histone deacetylase 3 protein. This gene is located adjacent to the ocular albinism gene and it is thought to be involved in the pathogenesis of the ocular albinism with late-onset sensorineural deafness phenotype. Four transcript variants encoding two different isoforms have been found for this gene. This gene is highly similar to the Y chromosome TBL1Y gene. [provided by RefSeq]
Sequence: MSITSDEVNFLVYRYLQESGFSHSAFTFGIESHISQSNINGTLVPPAALISILQKGLQYVEAEISINEDGTVFDGRPIESLSLIDAVMPDVVQTRQQAFREKLAQQQASAAAAAAAATAAATAATTTSAGVSHQNPSKNREATVNGEENRAHSVNNHAKPMEIDGEVEIPSSKATVLRGHESEVFICAWNPVSDLLASGSGDSTARIWNLNENSNGGSTQLVLRHCIREGGHDVPSNKDVTSLDWNTNGTLLATGSYDGFARIWTEDGNLASTLGQHKGPIFALKWNRKGNYILSAGVDKTTIIWDAHTGEAKQQFPFHSAPALDVDWQNNTTFASCSTDMCIHVCRLGCDRPVKTFQGHTNEVNAIKWDPSGMLLASCSDDMTLKIWSMKQEVCIHDLQAHNKEIYTIKWSPTGPATSNPNSNIMLASASFDSTVRLWDIERGVCTHTLTKHQEPVYSVAFSPDGKYLASGSFDKCVHIWNTQSGNLVHSYRGTGGIFEVCWNARGDKVGASASDGSVCVLDLRKSpezifikation
AAH32708.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
83.4kDa | |
Glutathione Sepharose 4 Fast Flow | |
MSITSDEVNFLVYRYLQESGFSHSAFTFGIESHISQSNINGTLVPPAALISILQKGLQYVEAEISINEDGTVFDGRPIESLSLIDAVMPDVVQTRQQAFREKLAQQQASAAAAAAAATAAATAATTTSAGVSHQNPSKNREATVNGEENRAHSVNNHAKPMEIDGEVEIPSSKATVLRGHESEVFICAWNPVSDLLASGSGDSTARIWNLNENSNGGSTQLVLRHCIREGGHDVPSNKDVTSLDWNTNGTLLATGSYDGFARIWTEDGNLASTLGQHKGPIFALKWNRKGNYILSAGVDKTTIIWDAHTGEAKQQFPFHSAPALDVDWQNNTTFASCSTDMCIHVCRLGCDRPVKTFQGHTNEVNAIKWDPSGMLLASCSDDMTLKIWSMKQEVCIHDLQAHNKEIYTIKWSPTGPATSNPNSNIMLASASFDSTVRLWDIERGVCTHTLTKHQEPVYSVAFSPDGKYLASGSFDKCVHIWNTQSGNLVHSYRGTGGIFEVCWNARGDKVGASASDGSVCVLDLRK | |
RUO | |
TBL1X | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
6907 | |
TBL1X (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
EBI/SMAP55/TBL1 | |
TBL1X | |
Recombinant | |
wheat germ expression system |