missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human STK11IP Partial ORF (NP_443134, 54 a.a. - 152 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marke: Abnova™ H00114790-Q01.25ug
Additional Details : Gewicht : 0.00010kg
Beschreibung
Sequence: HVFELHLGPWGPGQTGFVALPSHPADSPVILQLQFLFDVLQKTLSLKLVHVAGPGPTGPIKIFPFKSLRHLELRGVPLHCLHGLRGIYSQLETLICSRSSpezifikation
NP_443134 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.63kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
HVFELHLGPWGPGQTGFVALPSHPADSPVILQLQFLFDVLQKTLSLKLVHVAGPGPTGPIKIFPFKSLRHLELRGVPLHCLHGLRGIYSQLETLICSRS | |
RUO | |
STK11IP | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
114790 | |
STK11IP (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
KIAA1898/LIP1/LKB1IP/STK11IP1 | |
STK11IP | |
Recombinant | |
wheat germ expression system |