Learn More
ACROBiosystems Human Stathmin 1 / STMN1 Protein, His Tag
252.20 CHF - 1374.49 CHF
Spezifikation
Zugriffsnummer | NP_981946 |
---|---|
Molekulargewicht | 18.1 kDa |
Quelle | Human Stathmin 1, His Tag (ST1-H5149) is expressed from E.coli cells. It contains AA Ala 2 - Asp 149 (Accession # NP_981946). |
Immunogen | Ala 2 - Asp 149 |
Lagerungsbedingungen | -20°C |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
15817727
|
ACROBiosystems
ST1-H5149-100UG |
100 μg |
252.20 CHF
100 Mikrogramm |
Verfügbar ab: 15-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
15827727
|
ACROBiosystems
ST1-H5149-1MG |
1 mg |
1374.49 CHF
1 Milligramm |
Verfügbar ab: 15-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
Beschreibung
Stathmin (STMN1), a member of the stathmin family, is also known as Leukemia-associated phosphoprotein p18, Oncoprotein 18 (Op18), Phosphoprotein p19 (pp19), Protein Pr22 and pp17, which contains one SLD (stathmin-like) domain. The function of STMN1 as an important regulatory protein of microtubule dynamics has been well-characterized. Stathmin (STMN1) interacts with two molecules of dimeric α,β-tubulin to form a tight ternary complex called the T2S complex.One mole of STMN1 binds to two moles of tubulin dimers through the stathmin-like domain (SLD).When STMN1 sequesters tubulin into the T2S complex, tubulin becomes non-polymerizable. Without tubulin polymerization, there is no microtubule assembly.STMN1 also promotes microtubule disassembly by acting directly on the microtubule ends.
This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 18.1 kDa. The protein migrates as 18 kDa under reducing (R) condition (SDS-PAGE).Spezifikation
NP_981946 | |
Human Stathmin 1, His Tag (ST1-H5149) is expressed from E.coli cells. It contains AA Ala 2 - Asp 149 (Accession # NP_981946). | |
-20°C | |
Stathmin 1 | |
Human | |
PBS, pH7.4 | |
E.coli | |
Other Recombinant Proteins |
18.1 kDa | |
Ala 2 - Asp 149 | |
STMN1,Stathmin,C1orf215,LAP18,OP18 | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>92% as determined by SDS-PAGE |