Learn More
Abnova™ Human STAB1 Partial ORF (NP_055951, 1804 a.a. - 1902 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marke: Abnova™ H00023166-Q01.10ug
Additional Details : Gewicht : 0.00010kg
Beschreibung
This gene encodes a large, transmembrane receptor protein which may function in angiogenesis, lymphocyte homing, cell adhesion, or receptor scavenging. The protein contains 7 fasciclin, 16 epidermal growth factor (EGF)-like, and 2 laminin-type EGF-like domains as well as a C-type lectin-like hyaluronan-binding Link module. The protein is primarily expressed on sinusoidal endothelial cells of liver, spleen, and lymph node. The receptor has been shown to endocytose ligands such as low density lipoprotein, Gram-positive and Gram-negative bacteria, and advanced glycosylation end products. Supporting its possible role as a scavenger receptor, the protein rapidly cycles between the plasma membrane and early endosomes. [provided by RefSeq]
Sequence: EALASDLPNLGPLRTMHGTPISFSCSRTRAGELMVGEDDARIVQRHLPFEGGLAYGIDQLLEPPGLGARCDHFETRPLRLNTCSICGLEPPCPEGSQEQSpezifikation
NP_055951 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.63kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
EALASDLPNLGPLRTMHGTPISFSCSRTRAGELMVGEDDARIVQRHLPFEGGLAYGIDQLLEPPGLGARCDHFETRPLRLNTCSICGLEPPCPEGSQEQ | |
RUO | |
STAB1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
23166 | |
STAB1 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CLEVER-1/FEEL-1/FELE-1/FEX1/KIAA0246/STAB-1 | |
STAB1 | |
Recombinant | |
wheat germ expression system |