missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human ST13 Full-length ORF (AAH15317, 1 a.a. - 58 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
361.00 CHF - 547.00 CHF
Spezifikation
Zugriffsnummer | AAH15317 |
---|---|
Zur Verwendung mit (Anwendung) | Antibody Production, Protein Array, ELISA, Western Blot |
Zusammensetzung | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gen-ID (Entrez) | 6767 |
Molekulargewicht | 32.12kDa |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
16141052
|
Abnova™
H00006767-P01.25UG |
25 ug |
547.00 CHF
25 Mikrogramm |
Verfügbar ab: 03-06-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
16131052
|
Abnova™
H00006767-P01.10UG |
10 ug |
361.00 CHF
10 Mikrogramm |
Verfügbar ab: 03-06-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
Beschreibung
The protein encoded by this gene is an adaptor protein that mediates the association of the heat shock proteins HSP70 and HSP90. This protein has been shown to be involved in the assembly process of glucocorticoid receptor, which requires the assistance of multiple molecular chaperones. The expression of this gene is reported to be downregulated in colorectal carcinoma tissue suggesting that is a candidate tumor suppressor gene. [provided by RefSeq]
Sequence: MLFKSFKNTHHINLHLSLCVLLLMCRVLLSRNCQCVLGLTQEQFLLDSLFDLFNLIIFSpezifikation
AAH15317 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
32.12kDa | |
Glutathione Sepharose 4 Fast Flow | |
MLFKSFKNTHHINLHLSLCVLLLMCRVLLSRNCQCVLGLTQEQFLLDSLFDLFNLIIF | |
RUO | |
ST13 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
6767 | |
ST13 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
AAG2/FAM10A1/FAM10A4/FLJ27260/HIP/HOP/HSPABP/HSPABP1/MGC129952/P48/PRO0786/SNC6 | |
ST13 | |
Recombinant | |
wheat germ expression system |