Learn More
Abnova™ Human SSX1 Full-length ORF (NP_005626.1, 1 a.a. - 188 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marke: Abnova™ H00006756-P01.25ug
Additional Details : Gewicht : 0.00010kg
Beschreibung
The product of this gene belongs to the family of highly homologous synovial sarcoma X (SSX) breakpoint proteins. These proteins may function as transcriptional repressors. They are also capable of eliciting spontaneously humoral and cellular immune responses in cancer patients, and are potentially useful targets in cancer vaccine-based immunotherapy. SSX1, SSX2 and SSX4 genes have been involved in the t(X;18) translocation characteristically found in all synovial sarcomas. This translocation results in the fusion of the synovial sarcoma translocation gene on chromosome 18 to one of the SSX genes on chromosome X. The encoded hybrid proteins are probably responsible for transforming activity. [provided by RefSeq]
Sequence: MNGDDTFAKRPRDDAKASEKRSKAFDDIATYFSKKEWKKMKYSEKISYVYMKRNYKAMTKLGFKVTLPPFMCNKQATDFQGNDFDNDHNRRIQVEHPQMTFGRLHRIIPKIMPKKPAEDENDSKGVSEASGPQNDGKQLHPPGKANISEKINKRSGPKRGKHAWTHRLRERKQLVIYEEISDPEEDDESpezifikation
NP_005626.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
48.3kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
MGC150425/MGC5162/SSRC | |
SSX1 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
6756 | |
SSX1 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MNGDDTFAKRPRDDAKASEKRSKAFDDIATYFSKKEWKKMKYSEKISYVYMKRNYKAMTKLGFKVTLPPFMCNKQATDFQGNDFDNDHNRRIQVEHPQMTFGRLHRIIPKIMPKKPAEDENDSKGVSEASGPQNDGKQLHPPGKANISEKINKRSGPKRGKHAWTHRLRERKQLVIYEEISDPEEDDE | |
RUO | |
SSX1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |