missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human SRPK1 Partial ORF (AAH38292, 371 a.a. - 470 a.a.) Recombinant Protein with GST-tag at N-terminal
Click to view available options
Menge:
10 ug
25 ug
Packungsgröße:
10 Mikrogramm
25 Mikrogramm
Beschreibung
This gene encodes a serine/arginine protein kinase specific for the SR (serine/arginine-rich domain) family of splicing factors. The protein localizes to the nucleus and the cytoplasm. It is thought to play a role in regulation of both constitutive and alternative splicing by regulating intracellular localization of splicing factors. A second alternatively spliced transcript variant for this gene has been described, but its full length nature has not been determined. [provided by RefSeq]
Sequence: NETLRHKEDLHNANDCDVQNLNQESSFLSSQNGDSSTSQETDSCTPITSEVSDTMVCQSSSTVGQSFSEQHISQLQESIRAEIPCEDEQEQEHNGPLDNK
Spezifikation
Spezifikation
Zugriffsnummer | AAH38292 |
Zur Verwendung mit (Anwendung) | Antibody Production, ELISA, Protein Array, Western Blot |
Zusammensetzung | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gen-ID (Entrez) | 6732 |
Molekulargewicht | 36.63kDa |
Name | SRPK1 (Human) Recombinant Protein (Q01) |
Qualitätskontrollen | 12.5% SDS-PAGE Stained with Coomassie Blue. |
Menge | 25 ug |
Immunogen | NETLRHKEDLHNANDCDVQNLNQESSFLSSQNGDSSTSQETDSCTPITSEVSDTMVCQSSSTVGQSFSEQHISQLQESIRAEIPCEDEQEQEHNGPLDNK |
Lagerungsbedingungen | Store at -80°C. Aliquot to avoid repeated freezing and thawing. |
Mehr anzeigen |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Abnova™ Human SRPK1 Partial ORF (AAH38292, 371 a.a. - 470 a.a.) Recombinant Protein with GST-tag at N-terminal >
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur