missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human SPINT1 Full-length ORF (NP_001027539.1, 1 a.a. - 513 a.a.) Recombinant Protein with GST-tag at N-terminal
Click to view available options
Menge:
10 ug
25 ug
Packungsgröße:
10 Mikrogramm
25 Mikrogramm
Beschreibung
The protein encoded by this gene is a member of the Kunitz family of serine protease inhibitors. The protein is a potent inhibitor specific for HGF activator and is thought to be involved in the regulation of the proteolytic activation of HGF in injured tissues. Alternative splicing results in multiple variants encoding different isoforms. [provided by RefSeq]
Sequence: MAPARTMARARLAPAGIPAVALWLLCTLGLQGTQAGPPPAPPGLPAGADCLNSFTAGVPGFVLDTNASVSNGATFLESPTVRRGWDCVRACCTTQNCNLALVELQPDRGEDAIAACFLINCLYEQNFVCKFAPREGFINYLTREVYRSYRQLRTQGFGGSGIPKAWAGIDLKVQPQEPLVLKDVENTDWRLLRGDTDVRVERKDPNQVELWGLKEGTYLFQLTVTSSDHPEDTANVTVTVLSTKQTEDYCLASNKVGRCRGSFPRWYYDPTEQICKSFVYGGCLGNKNNYLREEECILACRGVQGPSMERRHPVCSGTCQPTQFRCSNGCCIDSFLECDDTPNCPDASDEAACEKYTSGFDELQRIHFPSDKGHCVDLPDTGLCKESIPRWYYNPFSEHCARFTYGGCYGNKNNFEEEQQCLESCRGISKKDVFGLRREIPIPSTGSVEMAVAVFLVICIVVVVAILGYCFFKNQRKDFHGHHHHPPPTPASSTVSTTEDTEHLVYNHTTRPL
Spezifikation
Spezifikation
Zugriffsnummer | NP_001027539.1 |
Zur Verwendung mit (Anwendung) | Antibody Production, Protein Array, ELISA, Western Blot |
Zusammensetzung | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gen-ID (Entrez) | 6692 |
Molekulargewicht | 83.3kDa |
Name | SPINT1 (Human) Recombinant Protein (P01) |
Reinigungsverfahren | Glutathione Sepharose 4 Fast Flow |
Qualitätskontrollen | 12.5% SDS-PAGE Stained with Coomassie Blue. |
Menge | 10 ug |
Immunogen | MAPARTMARARLAPAGIPAVALWLLCTLGLQGTQAGPPPAPPGLPAGADCLNSFTAGVPGFVLDTNASVSNGATFLESPTVRRGWDCVRACCTTQNCNLALVELQPDRGEDAIAACFLINCLYEQNFVCKFAPREGFINYLTREVYRSYRQLRTQGFGGSGIPKAWAGIDLKVQPQEPLVLKDVENTDWRLLRGDTDVRVERKDPNQVELWGLKEGTYLFQLTVTSSDHPEDTANVTVTVLSTKQTEDYCLASNKVGRCRGSFPRWYYDPTEQICKSFVYGGCLGNKNNYLREEECILACRGVQGPSMERRHPVCSGTCQPTQFRCSNGCCIDSFLECDDTPNCPDASDEAACEKYTSGFDELQRIHFPSDKGHCVDLPDTGLCKESIPRWYYNPFSEHCARFTYGGCYGNKNNFEEEQQCLESCRGISKKDVFGLRREIPIPSTGSVEMAVAVFLVICIVVVVAILGYCFFKNQRKDFHGHHHHPPPTPASSTVSTTEDTEHLVYNHTTRPL |
Mehr anzeigen |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
Abnova™ Human SPINT1 Full-length ORF (NP_001027539.1, 1 a.a. - 513 a.a.) Recombinant Protein with GST-tag at N-terminal >
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido