missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human SPINT1 Full-length ORF (NP_001027539.1, 1 a.a. - 513 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
361.00 CHF - 547.00 CHF
Spezifikation
Zugriffsnummer | NP_001027539.1 |
---|---|
Zur Verwendung mit (Anwendung) | Antibody Production, Protein Array, ELISA, Western Blot |
Zusammensetzung | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gen-ID (Entrez) | 6692 |
Molekulargewicht | 83.3kDa |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
16143128
|
Abnova™
H00006692-P01.25UG |
25 ug |
547.00 CHF
25 Mikrogramm |
Verfügbar ab: 31-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
16133128
|
Abnova™
H00006692-P01.10UG |
10 ug |
361.00 CHF
10 Mikrogramm |
Verfügbar ab: 31-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
Beschreibung
The protein encoded by this gene is a member of the Kunitz family of serine protease inhibitors. The protein is a potent inhibitor specific for HGF activator and is thought to be involved in the regulation of the proteolytic activation of HGF in injured tissues. Alternative splicing results in multiple variants encoding different isoforms. [provided by RefSeq]
Sequence: MAPARTMARARLAPAGIPAVALWLLCTLGLQGTQAGPPPAPPGLPAGADCLNSFTAGVPGFVLDTNASVSNGATFLESPTVRRGWDCVRACCTTQNCNLALVELQPDRGEDAIAACFLINCLYEQNFVCKFAPREGFINYLTREVYRSYRQLRTQGFGGSGIPKAWAGIDLKVQPQEPLVLKDVENTDWRLLRGDTDVRVERKDPNQVELWGLKEGTYLFQLTVTSSDHPEDTANVTVTVLSTKQTEDYCLASNKVGRCRGSFPRWYYDPTEQICKSFVYGGCLGNKNNYLREEECILACRGVQGPSMERRHPVCSGTCQPTQFRCSNGCCIDSFLECDDTPNCPDASDEAACEKYTSGFDELQRIHFPSDKGHCVDLPDTGLCKESIPRWYYNPFSEHCARFTYGGCYGNKNNFEEEQQCLESCRGISKKDVFGLRREIPIPSTGSVEMAVAVFLVICIVVVVAILGYCFFKNQRKDFHGHHHHPPPTPASSTVSTTEDTEHLVYNHTTRPLSpezifikation
NP_001027539.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
83.3kDa | |
Glutathione Sepharose 4 Fast Flow | |
MAPARTMARARLAPAGIPAVALWLLCTLGLQGTQAGPPPAPPGLPAGADCLNSFTAGVPGFVLDTNASVSNGATFLESPTVRRGWDCVRACCTTQNCNLALVELQPDRGEDAIAACFLINCLYEQNFVCKFAPREGFINYLTREVYRSYRQLRTQGFGGSGIPKAWAGIDLKVQPQEPLVLKDVENTDWRLLRGDTDVRVERKDPNQVELWGLKEGTYLFQLTVTSSDHPEDTANVTVTVLSTKQTEDYCLASNKVGRCRGSFPRWYYDPTEQICKSFVYGGCLGNKNNYLREEECILACRGVQGPSMERRHPVCSGTCQPTQFRCSNGCCIDSFLECDDTPNCPDASDEAACEKYTSGFDELQRIHFPSDKGHCVDLPDTGLCKESIPRWYYNPFSEHCARFTYGGCYGNKNNFEEEQQCLESCRGISKKDVFGLRREIPIPSTGSVEMAVAVFLVICIVVVVAILGYCFFKNQRKDFHGHHHHPPPTPASSTVSTTEDTEHLVYNHTTRPL | |
RUO | |
SPINT1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
6692 | |
SPINT1 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
HAI/HAI1/MANSC2 | |
SPINT1 | |
Recombinant | |
wheat germ expression system |