missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human SPAG7 Full-length ORF (AAH11934.1, 1 a.a. - 227 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marke: Abnova™ H00009552-P01.10ug
Additional Details : Gewicht : 0.00010kg
Beschreibung
Sequence: MADLLGSILSSMEKPPSLGDQETRRKAREQAARLKKLQEQEKQQKVEFRKRMEKEVSDFIQDSGQIKKKFQPMNKIERSILHDVVEVAGLTSFSFGEDDDCRYVMIFKKEFAPSDEELDSYRRGEEWDPQKAEEKRKLKELAQRQEEEAAQQGPVVVSPASDYKDKYSHLIGKGAAKDAAHMLQANKTYGCVPVANKRDTRSIEEAMNEIRAKKRLRQSGEELPPTSSpezifikation
AAH11934.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
52.4kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ACRP/FSA-1/MGC20134 | |
SPAG7 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
9552 | |
SPAG7 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MADLLGSILSSMEKPPSLGDQETRRKAREQAARLKKLQEQEKQQKVEFRKRMEKEVSDFIQDSGQIKKKFQPMNKIERSILHDVVEVAGLTSFSFGEDDDCRYVMIFKKEFAPSDEELDSYRRGEEWDPQKAEEKRKLKELAQRQEEEAAQQGPVVVSPASDYKDKYSHLIGKGAAKDAAHMLQANKTYGCVPVANKRDTRSIEEAMNEIRAKKRLRQSGEELPPTS | |
RUO | |
SPAG7 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |