missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human SLC2A10 Partial ORF (NP_110404, 325 a.a. - 382 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marke: Abnova™ H00081031-Q01.10ug
Additional Details : Gewicht : 0.00010kg
Beschreibung
SLC2A10 is a member of the facilitative glucose transporter family, which plays a significant role in maintaining glucose homeostasis.[supplied by OMIM]
Sequence: SGPSCLAVPNATGQTGLPGDSGLLQDSSLPPIPRTNEDQREPILSTAKKTKPHPRSGDSpezifikation
NP_110404 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
32.12kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
SGPSCLAVPNATGQTGLPGDSGLLQDSSLPPIPRTNEDQREPILSTAKKTKPHPRSGD | |
RUO | |
SLC2A10 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
81031 | |
SLC2A10 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ATS/GLUT10/MGC126706 | |
SLC2A10 | |
Recombinant | |
wheat germ expression system |