Learn More
ACROBiosystems Human SLAMF1 / SLAM / CD150 Protein, His Tag
Marke: ACROBiosystems SL1-H52H0-1MG
Additional Details : Gewicht : 0.00001kg
Beschreibung
Self-ligand receptor of the signaling lymphocytic activation molecule (SLAM) family, also known as CD150, is the prototypic member of the SLAM subfamily of the CD2 protein family.SLAM receptors triggered by homo- or heterotypic cell-cell interactions are modulating the activation and differentiation of a wide variety of immune cells and thus are involved in the regulation and interconnection of both innate and adaptive immune response. Activities are controlled by presence or absence of small cytoplasmic adapter proteins, SH2D1A/SAP and/or SH2D1B/EAT-2. SLAMF1-induced signal-transduction events in T-lymphocytes are different from those in B-cells. The type I transmembrane glycoprotein Signaling Lymphocytic Activation Molecule (SLAM).
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 26.2 kDa. The protein migrates as 42-60 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spezifikation
NP_003028.1 | |
1 mg | |
Ala 21 - Pro 237 | |
SLAMF1,CD150,CDw150,SLAM,IPO-3 | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |
26.2 kDa | |
Human SLAMF1, His Tag (SL1-H52H0) is expressed from human 293 cells (HEK293). It contains AA Ala 21 - Pro 237 (Accession # Q13291-1). | |
-20°C | |
SLAMF1 | |
Human | |
PBS, pH7.4 | |
HEK293 | |
Immune Checkpoint Proteins |