missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human SGIP1 Full-length ORF (AAH40516.1, 1 a.a. - 859 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00084251-P01.10ug

Additional Details : Gewicht : 0.00010kg

 View more versions of this product

Product Code. 16125733

  • 379.00 CHF / 10 Mikrogramm
Verfügbar ab: 17-01-2024
Sie sich ein, um den Lagerbestand zu sehen
Add to basket

Description

Description

SGIP1 functions as an endocytic protein that affects signaling by receptors in neuronal systems involved in energy homeostasis via its interaction with endophilins (see SH3GL3; MIM 603362) (Trevaskis et al., 2005 [PubMed 15919751] and Uezu et al., 2007 [PubMed 17626015]).[supplied by OMIM]

Sequence: MMEGLKKRTRKAFGIRKKEKDTDSTGSPDRDGIQGKKKTQKTQLLLTSFFWLRALSLTLSQKKSNGAPNGFYAEIDWERYNSPELDEEGYSIRPEEPGSTKGKHFYSSSESEEEEESHKKFNIKIKPLQSKDILKNAATVDELKASIGNIALSPSPVRKSPRRSQGAIKRNLSSEEVARPRRSTPTPELISKKPPDDTTALAPLFGPPLESAFDEQKTEVLLDQPEIWGSGQPINPSMESPKLTRPFPTGTPPPLPPKNVPATPPRTGSPLTIGPGNDQSATEVKIEKLPSINDLDSIFGPVLSPKSVAVNAEEKWVHFSDTSPEHVTPELTPREKVVSPPATPDNPADSPAPGPLGPPGPTGPPGPPGPPRNVLSPLNLEEVQKKVAEQTFIKDDYLETISSPKDFGLGQRATPPPPPPPTYRTVVSSPGPGSGPGPGTTSDGKTEAQRYQVICPSLQAGGNELDSYSGASSPARPATPLVPCRSTTPPPPPPRPPSRPKLPPGKPGVGDVSRPFSPPIHSSSPPPIAPLARAESTSSISSTNSLSAATTPTVENEQPSLVWFDRGKFYLTFEGSSRGPSPLTMGAQDTLPVAAAFTETVNAYFEGADPSKCIVKITGEMVLSFPAGITRHFANNPSPAALTFRVINFSRLEHVLPNPQLLCCDNTQNDANTKEFWVNMPNLMTHLKKVSEQKPQATYYNVDMLKYQVSAQGIQSTPLNLAVNWRCEPSSTDLRIDYKYNTDAMTTAVALNNVQFLVPIDGGVTKLQAVLPPAVWNAEQQRILWKIPDISQKSENGGVGSLLARFQLSEGPSKPSPLVVQFTSEGSTLSGCDIELVGAGYRFSLIKKRFAAGKYLADN
Specifications

Specifications

AAH40516.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
119.1kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
DKFZp686A16142/DKFZp761D221/FLJ33378/FLJ43054
SGIP1
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
84251
SGIP1 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MMEGLKKRTRKAFGIRKKEKDTDSTGSPDRDGIQGKKKTQKTQLLLTSFFWLRALSLTLSQKKSNGAPNGFYAEIDWERYNSPELDEEGYSIRPEEPGSTKGKHFYSSSESEEEEESHKKFNIKIKPLQSKDILKNAATVDELKASIGNIALSPSPVRKSPRRSQGAIKRNLSSEEVARPRRSTPTPELISKKPPDDTTALAPLFGPPLESAFDEQKTEVLLDQPEIWGSGQPINPSMESPKLTRPFPTGTPPPLPPKNVPATPPRTGSPLTIGPGNDQSATEVKIEKLPSINDLDSIFGPVLSPKSVAVNAEEKWVHFSDTSPEHVTPELTPREKVVSPPATPDNPADSPAPGPLGPPGPTGPPGPPGPPRNVLSPLNLEEVQKKVAEQTFIKDDYLETISSPKDFGLGQRATPPPPPPPTYRTVVSSPGPGSGPGPGTTSDGKTEAQRYQVICPSLQAGGNELDSYSGASSPARPATPLVPCRSTTPPPPPPRPPSRPKLPPGKPGVGDVSRPFSPPIHSSSPPPIAPLARAESTSSISSTNSLSAATTPTVENEQPSLVWFDRGKFYLTFEGSSRGPSPLTMGAQDTLPVAAAFTETVNAYFEGADPSKCIVKITGEMVLSFPAGITRHFANNPSPAALTFRVINFSRLEHVLPNPQLLCCDNTQNDANTKEFWVNMPNLMTHLKKVSEQKPQATYYNVDMLKYQVSAQGIQSTPLNLAVNWRCEPSSTDLRIDYKYNTDAMTTAVALNNVQFLVPIDGGVTKLQAVLPPAVWNAEQQRILWKIPDISQKSENGGVGSLLARFQLSEGPSKPSPLVVQFTSEGSTLSGCDIELVGAGYRFSLIKKRFAAGKYLADN
RUO
SGIP1
Wheat Germ (in vitro)
GST
Liquid
Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions