Learn More
Abnova™ Human SERPINC1 native Protein from human plasma
Beschreibung
The protein encoded by this gene is a plasma protease inhibitor and a member of the serpin superfamily. This protein inhibits thrombin as well as other activated serine proteases of the coagulation system, and it regulates the blood coagulation cascade. The protein includes two functional domains: the heparin binding-domain at the N-terminus of the mature protein, and the reactive site domain at the C-terminus. The inhibitory activity is enhanced by the presence of heparin. More than 120 mutations have been identified for this gene, many of which are known to cause antithrombin-III deficiency. [provided by RefSeq]
Spezifikation
Spezifikation
| Konzentration | 8.3 mg/mL |
| Zur Verwendung mit (Anwendung) | SDS-PAGE |
| Zusammensetzung | In 0.05M Tris-HCl, 0.1M NaCl, pH 7.4 |
| Gen-ID (Entrez) | 462 |
| Molekulargewicht | 58kDa |
| Name | SERPINC1 (Human) Native Protein |
| Vorbereitungsmethode | Native protein purified from human plasma |
| Reinigungsverfahren | Multi-step chromatography |
| Menge | 1 mg |
| Lagerungsbedingungen | Store at -80°C. Aliquot to avoid repeated freezing and thawing. |
| Mehr anzeigen |
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.