Learn More
ACROBiosystems Human Serpin F1 / PEDF Protein, His Tag
Marke: ACROBiosystems SE1-H5221-1MG
Additional Details : Gewicht : 0.00001kg
Beschreibung
erpin F1 (SERPINF1) is also known as Pigment epithelium-derived factor (PEDF), Cell proliferation-inducing gene 35 protein (PIG35). Serpin F1 belongs to the serpin family. Serpin F1 is expressed in quiescent cells. PEDF has a variety of functions including antiangiogenic, antitumorigenic, and neurotrophic properties. Endothelial cell migration is inhibited by SERPINF1/ PEDF. PEDF / SERPINF1 suppresses retinal neovascularization and endothelial cell proliferation. PEDF is also responsible for apoptosis of endothelial cells either through the p38 MAPK pathway or through the FAS/FASL pathway. PEDF also displays neurotrophic functions.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 45.2 kDa. The protein migrates as 45-60 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spezifikation
NP_002606 | |
1 mg | |
Gln 20 - Pro 418 | |
SERPINF1,Serpin F1,PEDF,PIG35,EPC-1 | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |
45.2 kDa | |
Human Serpin F1, His Tag (SE1-H5221) is expressed from human 293 cells (HEK293). It contains AA Gln 20 - Pro 418 (Accession # NP_002606). | |
-20°C | |
Serpin F1 | |
Human | |
PBS, pH7.4 | |
HEK293 | |
Other Recombinant Proteins |