missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACROBiosystems Human Serpin A3 Protein, His Tag
403.52 CHF - 3076.85 CHF
Spezifikation
Zugriffsnummer | AAH03559 |
---|---|
Molekulargewicht | 46.6 kDa |
Quelle | Human Serpin A3, His Tag (SE3-H5221) is expressed from human 293 cells (HEK293). It contains AA His 24 - Ala 423 (Accession # AAH03559). |
Immunogen | His 24 - Ala 423 |
Lagerungsbedingungen | -20°C |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
15807677
|
ACROBiosystems
SE3-H5221-1MG |
1 mg |
3076.85 CHF
1 Milligramm |
Verfügbar ab:
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
15817677
|
ACROBiosystems
SE3-H5221-50UG |
50 μg |
403.52 CHF
50 Mikrogramm |
Verfügbar ab:
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
Beschreibung
Serpin A3 is also known as Alpha-1-antichymotrypsin (ACT or AACT), Cell growth-inhibiting gene 24/25 protein, which belongs to the serpin family. Serpin A3 can bind DNA. Although its physiological function is unclear, it can inhibit neutrophil cathepsin G and mast cell chymase, both of which can convert angiotensin-1 to the active angiotensin-2.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 46.6 kDa. The protein migrates as 50-63 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spezifikation
AAH03559 | |
Human Serpin A3, His Tag (SE3-H5221) is expressed from human 293 cells (HEK293). It contains AA His 24 - Ala 423 (Accession # AAH03559). | |
-20°C | |
Serpin A3 | |
Human | |
PBS, pH7.4 | |
HEK293 | |
Other Recombinant Proteins |
46.6 kDa | |
His 24 - Ala 423 | |
SERPINA3,ACT,AACT,GIG24,GIG25 | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |