Learn More
Abnova™ Human SEPHS2 Full-length ORF (NP_036380.2, 1 a.a. - 59 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marke: Abnova™ H00022928-P01.25ug
Additional Details : Gewicht : 0.00010kg
Beschreibung
This gene encodes an enzyme that synthesizes selenophosphate from selenide and ATP. Selenophosphate is the selenium donor used to synthesize selenocysteine, which is co-translationally incorporated into selenoproteins at in-frame UGA codons. This protein itself contains a selenocysteine residue in its predicted active site. The 3' UTR of the gene has a stem-loop secondary structure called a selenocysteine insertion sequence (SECIS) element, which allows UGA to direct the incorporation of selenocysteine rather than signal a translational stop. Alternatively spliced transcripts have been identified, but their biological validity has not been determined. [provided by RefSeq]
Sequence: MAEASATGACGEAMAAAEGSSGPAGLTLGRSFSNYRPFEPQALGLSPSWRLTGFSGMKGSpezifikation
NP_036380.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
32.3kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
SPS2 | |
SEPHS2 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
22928 | |
SEPHS2 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MAEASATGACGEAMAAAEGSSGPAGLTLGRSFSNYRPFEPQALGLSPSWRLTGFSGMKG | |
RUO | |
SEPHS2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |