missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human SEC16B Full-length ORF (AAH09106.1, 1 a.a. - 625 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marke:  Abnova™ H00089866-P01.25ug

Additional Details : Gewicht : 0.00010kg

 Lernen Sie weitere Varianten dieses

Artikelnummer. 16126413

  • 547.00 CHF / 25 Mikrogramm
Verfügbar ab: 09-01-2024
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

SEC16B is a mammalian homolog of S. cerevisiae Sec16 that is required for organization of transitional endoplasmic reticulum (ER) sites and protein export (Bhattacharyya and Glick, 2007 [PubMed 17192411]).[supplied by OMIM]

Sequence: MELWAPQRLPQTRGKATAPSKDPDRGFRRDGHHRPVPHSWHNGERFHQWQDNRGSPQPQQEPRADHQQQPHYASRPGDWHQPVSGVDYYEGGYRNQLYSRPGYENSYQSYQSPTMREEYAYGSYYYHGHPQWLQEERVPRQRSPYIWHEDYREQKYLDEHHYENQHSPFGTNSETHFQSNSRNPCKDSPASNSGQEWPGELFPGSLLAEAQKNKPSLASESNLLQQRESGLSSSSYELSQYIRDAPERDDPPASAAWSPVQAEDVSSAGPKAPMKFYIPHVPVSFGPGGQLVRVGPSSPTDGQAALVELHSMEVILNDSEEQEEMRSFSGPLIREDVHKVDIMTFCQQKAAQSCKSETLGSRDSALLWQLLVLLCRQNGSMVGSDIAELLMQDCKKLEKYKRQPPVANLINLTDEDWPVLSSGTPNLLTGEIPPSVETPAQIVEKFTRLLYYGRKKEALEWAMKNHLWGHALFLSSKMDPQTYSWVMSGFTSTLALNDPLQTLFQLMSGRIPQAATCCGEKQWGDWRPHLAVILSNQAGDPELYQRAIVAIGDTLAGKGLVEAAHFCYLMAHVPFGHYTVKTDHLVLLGSSHRYATWEKGNSKDIFQGTVLALVGFYGSSFHFLM
Spezifikation

Spezifikation

AAH09106.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
97.2kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
DKFZp686C2486/FLJ23871/FLJ25761/FLJ33652/FLJ36620/LZTR2/PGPR-p117/RGPR/SEC16S
SEC16B
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
89866
SEC16B (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MELWAPQRLPQTRGKATAPSKDPDRGFRRDGHHRPVPHSWHNGERFHQWQDNRGSPQPQQEPRADHQQQPHYASRPGDWHQPVSGVDYYEGGYRNQLYSRPGYENSYQSYQSPTMREEYAYGSYYYHGHPQWLQEERVPRQRSPYIWHEDYREQKYLDEHHYENQHSPFGTNSETHFQSNSRNPCKDSPASNSGQEWPGELFPGSLLAEAQKNKPSLASESNLLQQRESGLSSSSYELSQYIRDAPERDDPPASAAWSPVQAEDVSSAGPKAPMKFYIPHVPVSFGPGGQLVRVGPSSPTDGQAALVELHSMEVILNDSEEQEEMRSFSGPLIREDVHKVDIMTFCQQKAAQSCKSETLGSRDSALLWQLLVLLCRQNGSMVGSDIAELLMQDCKKLEKYKRQPPVANLINLTDEDWPVLSSGTPNLLTGEIPPSVETPAQIVEKFTRLLYYGRKKEALEWAMKNHLWGHALFLSSKMDPQTYSWVMSGFTSTLALNDPLQTLFQLMSGRIPQAATCCGEKQWGDWRPHLAVILSNQAGDPELYQRAIVAIGDTLAGKGLVEAAHFCYLMAHVPFGHYTVKTDHLVLLGSSHRYATWEKGNSKDIFQGTVLALVGFYGSSFHFLM
RUO
SEC16B
Wheat Germ (in vitro)
GST
Liquid
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote