missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human SAMHD1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Marke: R&D Systems MAB8120-SP
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
SAMHD1 Monoclonal specifically detects SAMHD1 in Human samples. It is validated for Western Blot, Simple Western.
Spezifikation
| SAMHD1 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL, Simple Western 10 ug/mL | |
| Q9Y3Z3 | |
| SAMHD1 | |
| E. coli-derived recombinant human SAMHD1 His206-Arg339 Accession # Q9Y3Z3 | |
| 25 μg | |
| Primary | |
| Detects human SAMHD1 in ELISAs. | |
| Human | |
| Purified |
| Western Blot | |
| 883335 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| AGS5, CHBL2, DCIP, DCIPMg11, Dendritic cell-derived IFNG-induced protein, EC 3.1.4.-, HDDC1, Monocyte protein 5, MOP5, MOP-5, MOP-5AGS5, SAM domain and HD domain 1, SAM domain and HD domain-containing protein 1, SBBI88 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 25939 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
For Research Use Only
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur