missing translation for 'onlineSavingsMsg'
Learn More

ACROBiosystems Human S100P / S100E Protein, Tag Free

312.65 CHF - 1638.26 CHF

Spezifikation

Zugriffsnummer AAH06819
Molekulargewicht 10.4 kDa
Quelle Human S100P, Tag Free (S1P-H5110) is expressed from E.coli cells. It contains AA Met 1 - Lys 95 (Accession # AAH06819).
Immunogen Met 1 - Lys 95
Lagerungsbedingungen -20°C
Mehr Spezifikationen ansehen

Produkte 2
Produktcode Marke Menge Preis Menge & Verfügbarkeit  
Produktcode Marke Menge Preis Menge & Verfügbarkeit  
15827647
Unterlagen ansehen Aktionsdetails
ACROBiosystems
S1P-H5110-1MG
1 mg
1638.26 CHF
1 Milligramm
Alternative Produkte anzeigen
Sparen Sie bis zu 
»
Verfügbar ab: 11-04-2024
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen
 
15817647
Unterlagen ansehen Aktionsdetails
ACROBiosystems
S1P-H5110-100UG
100 μg
312.65 CHF
100 Mikrogramm
Alternative Produkte anzeigen
Sparen Sie bis zu 
»
Verfügbar ab: 11-04-2024
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen
 
Beschreibung

Beschreibung

S100 calcium-binding protein P (S100P),a member of the S-100 family, is also known as protein S100-P, migration-inducing gene 9 protein (MIG9) and protein S100-E,which contains two EF-hand domains. As for subunit structure,S100P is homodimer and heterodimer with S100A1. S100P may function as calcium sensor and contribute to cellular calcium signaling. In a calcium-dependent manner, S100P functions by interacting with other proteins, such as EZR and PPP5C, and indirectly plays a role in physiological processes like the formation of microvilli in epithelial cells. Furthermore,S100P may stimulate cell proliferation in an autocrine manner via activation of the receptor for activated glycation end products (RAGE).

This protein carries no "e;tag"e;. The protein has a calculated MW of 10.4 kDa. The protein migrates as 10.4 kDa under reducing (R) condition (SDS-PAGE).
Spezifikation

Spezifikation

AAH06819
Human S100P, Tag Free (S1P-H5110) is expressed from E.coli cells. It contains AA Met 1 - Lys 95 (Accession # AAH06819).
-20°C
S100P
Human
50 mM Tris, 150 mM NaCl, pH8.0
E.coli
Bio-Markers and CD Antigens
10.4 kDa
Met 1 - Lys 95
S100P,S100E,MIG9
Unconjugated
Tag Free
Less than 1.0 EU / μg by the LAL method.
Powder
>95% as determined by SDS-PAGE
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Product Certifications