missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human S100B His-tag, Recombinant Protein
E. coli Recombinant Protein
Marke: Invitrogen™ A42515
Additional Details : Gewicht : 0.05000kg
Beschreibung
S100B is a member of the S100 family of proteins containing 2 EF-hand calcium-binding motifs. S100 proteins are localized in the cytoplasm and/or nucleus of a wide range of cells and are involved in the regulation of a number of cellular processes such as cell cycle progression and differentiation. S100B may function in neurite extension, proliferation of melanoma cells, stimulation of Ca2+ fluxes, inhibition of PKC-mediated phosphorylation, astrocytosis and axonal proliferation, and inhibition of microtubule assembly. Chromosomal rearrangements and altered expression of this gene have been implicated in several diseases, including Alzheimer's disease, Down's syndrome, epilepsy, amyotrophic lateral sclerosis, melanoma, and type I diabetes.Spezifikation
P04271 | |
PBS with 0.2 μg/mL BSA, 10% glycerol and no preservative; pH 7.5 | |
Human S100B His-tag | |
E. coli | |
RUO | |
S100B | |
Unconjugated | |
Liquid |
Control, ELISA, Functional Assay, Immunohistochemistry, Western Blot | |
6285 | |
50 μg | |
-80°C | |
AI850290; Bpb; hypothetical protein LOC525716; NEF; Protein S100-B; S100; S100 calcium binding protein B; S100 calcium-binding protein B; S100 calcium-binding protein beta (neural); S-100 calcium-binding protein beta subunit; S100 calcium-binding protein, beta (neural); S-100 calcium-binding protein, beta chain; S-100 protein beta chain; S-100 protein subunit beta; S100 protein, beta polypeptide; S100 protein, beta polypeptide, neural; S100b; S100-B; S100beta; S100P | |
Human | |
Recombinant | |
≥ 90% |