missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human RYK Partial ORF (NP_002949, 54 a.a. - 163 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

361.00 CHF - 547.00 CHF

Spezifikation

Zugriffsnummer NP_002949
Zur Verwendung mit (Anwendung) Antibody Production, ELISA, Protein Array, Western Blot
Zusammensetzung 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gen-ID (Entrez) 6259
Molekulargewicht 37.84kDa
Mehr Spezifikationen ansehen

Produkte 2
Produktcode Marke Menge Preis Menge & Verfügbarkeit  
Produktcode Marke Menge Preis Menge & Verfügbarkeit  
16134835
Unterlagen ansehen Aktionsdetails
Abnova™
H00006259-Q01.10UG
10 ug
361.00 CHF
10 Mikrogramm
Alternative Produkte anzeigen
Sparen Sie bis zu 
»
Verfügbar ab: 07-05-2024
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen
 
16144835
Unterlagen ansehen Aktionsdetails
Abnova™
H00006259-Q01.25UG
25 ug
547.00 CHF
25 Mikrogramm
Alternative Produkte anzeigen
Sparen Sie bis zu 
»
Verfügbar ab: 07-05-2024
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen
 
Beschreibung

Beschreibung

The protein encoded by this gene is an atypical member of the family of growth factor receptor protein tyrosine kinases, differing from other members at a number of conserved residues in the activation and nucleotide binding domains. This gene product belongs to a subfamily whose members do not appear to be regulated by phosphorylation in the activation segment. It has been suggested that mediation of biological activity by recruitment of a signaling-competent auxiliary protein may occur through an as yet uncharacterized mechanism. Two alternative splice variants have been identified, encoding distinct isoforms. [provided by RefSeq]

Sequence: ELQSASAGPSVSLYLSEDEVRRLIGLDAELYYVRNDLISHYALSFNLLVPSETNFLHFTWHAKSKVEYKLGFQVDNVLAMDMPQVNISVQGEVPRTLSVFRVELSCTGKV
Spezifikation

Spezifikation

NP_002949
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
37.84kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
D3S3195/JTK5/JTK5A/RYK1
RYK
Recombinant
wheat germ expression system
Antibody Production, ELISA, Protein Array, Western Blot
6259
RYK (Human) Recombinant Protein (Q01)
ELQSASAGPSVSLYLSEDEVRRLIGLDAELYYVRNDLISHYALSFNLLVPSETNFLHFTWHAKSKVEYKLGFQVDNVLAMDMPQVNISVQGEVPRTLSVFRVELSCTGKV
RUO
RYK
Wheat Germ (in vitro)
GST
Liquid
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Product Certifications