missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human RPS15 Full-length ORF (AAH64908, 1 a.a. - 145 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marke:  Abnova™ H00006209-P01.10ug

Additional Details : Gewicht : 0.00010kg

 Lernen Sie weitere Varianten dieses

Artikelnummer. 16120372

  • 361.00 CHF / 10 Mikrogramm
Verfügbar ab: 11-06-2024
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

Ribosomes, the organelles that catalyze protein synthesis, consist of a small 40S subunit and a large 60S subunit. Together these subunits are composed of 4 RNA species and approximately 80 structurally distinct proteins. This gene encodes a ribosomal protein that is a component of the 40S subunit. The protein belongs to the S19P family of ribosomal proteins. It is located in the cytoplasm. This gene has been found to be activated in various tumors, such as insulinomas, esophageal cancers, and colon cancers. As is typical for genes encoding ribosomal proteins, there are multiple processed pseudogenes of this gene dispersed through the genome. [provided by RefSeq]

Sequence: MAEVEQKKKRTFRKFTYRGVDLDQLLDMSYEQLMQLYSARQRRRLNRGLRRKQHSLLKRLRKAKKEAPPMEKPEVVKTHLRDMIILPEMVGSMVGVYNGKTFNQVEIKPEMIGHYLGEFSITYKPVKHGRPGIGATHSSRFIPLK
Spezifikation

Spezifikation

AAH64908
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
41.69kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
MGC111130/RIG
RPS15
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
6209
RPS15 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MAEVEQKKKRTFRKFTYRGVDLDQLLDMSYEQLMQLYSARQRRRLNRGLRRKQHSLLKRLRKAKKEAPPMEKPEVVKTHLRDMIILPEMVGSMVGVYNGKTFNQVEIKPEMIGHYLGEFSITYKPVKHGRPGIGATHSSRFIPLK
RUO
RPS15
Wheat Germ (in vitro)
GST
Liquid
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote