missing translation for 'onlineSavingsMsg'
Learn More
Abnova™ Human RPLP0 Full-length ORF (AAH08092, 1 a.a. - 317 a.a.) Recombinant Protein with GST-tag at N-terminal Artikelnummer.: 16130322
Nur noch undefined auf Lager
Zum Warenkorb hinzufügen

Abnova™ Human RPLP0 Full-length ORF (AAH08092, 1 a.a. - 317 a.a.) Recombinant Protein with GST-tag at N-terminal

Artikelnummer. 16130322
25 ug, 25 Mikrogramm
Click to view available options
Menge:
10 ug
25 ug
Packungsgröße:
10 Mikrogramm
25 Mikrogramm
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Artikelnummer. 16130322

Marke: Abnova™ H00006175P01.25ug

Nur noch undefined auf Lager
Zum Warenkorb hinzufügen
Nur noch undefined auf Lager
Zum Warenkorb hinzufügen
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Used for AP, Array, ELISA, WB-Re

Ribosomes, the organelles that catalyze protein synthesis, consist of a small 40S subunit and a large 60S subunit. Together these subunits are composed of 4 RNA species and approximately 80 structurally distinct proteins. This gene encodes a ribosomal protein that is a component of the 60S subunit. The protein, which is the functional equivalent of the E. coli L10 ribosomal protein, belongs to the L10P family of ribosomal proteins. It is a neutral phosphoprotein with a C-terminal end that is nearly identical to the C-terminal ends of the acidic ribosomal phosphoproteins P1 and P2. The P0 protein can interact with P1 and P2 to form a pentameric complex consisting of P1 and P2 dimers, and a P0 monomer. The protein is located in the cytoplasm. Transcript variants derived from alternative splicing exist; they encode the same protein. As is typical for genes encoding ribosomal proteins, there are multiple processed pseudogenes of this gene dispersed through the genome. [provided by RefSeq]

Sequence: MPREDRATWKSNYFLKIIQLLDDYPKCFIVGADNVGSKQMQQIRMSLRGKAVVLMGKNTMMRKAIRGHLENNPALEKLLPHIRGNVGFVFTKEDLTEIRDMLLANKVPAAARAGAIAPCEVTVPAQNTGLGPEKTSFFQALGITTKISRGTIEILSDVQLIKTGDKVGASEATLLNMLNISPFSFGLVIQQVFDNGSIYNPEVLDITEETLHSRFLEGVRNVASVCLQIGYPTVASVPHSIINGYKRVLALSVETDYTFPLAEKVKAFLADPSAFVAAAPVAAATTAAPAAAAAPAKVEAKEESEESDEDMGFGLFD

Spezifikation

Zugriffsnummer AAH08092
Zur Verwendung mit (Anwendung) Antibody Production, Protein Array, ELISA, Western Blot
Zusammensetzung 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gen-ID (Entrez) 6175
Molekulargewicht 60.61kDa
Name RPLP0 (Human) Recombinant Protein (P01)
Reinigungsverfahren Glutathione Sepharose 4 Fast Flow
Qualitätskontrollen 12.5% SDS-PAGE Stained with Coomassie Blue.
Menge 25 ug
Immunogen MPREDRATWKSNYFLKIIQLLDDYPKCFIVGADNVGSKQMQQIRMSLRGKAVVLMGKNTMMRKAIRGHLENNPALEKLLPHIRGNVGFVFTKEDLTEIRDMLLANKVPAAARAGAIAPCEVTVPAQNTGLGPEKTSFFQALGITTKISRGTIEILSDVQLIKTGDKVGASEATLLNMLNISPFSFGLVIQQVFDNGSIYNPEVLDITEETLHSRFLEGVRNVASVCLQIGYPTVASVPHSIINGYKRVLALSVETDYTFPLAEKVKAFLADPSAFVAAAPVAAATTAAPAAAAAPAKVEAKEESEESDEDMGFGLFD
Lagerungsbedingungen Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Kennzeichnung RUO
Gen-Alias L10E/MGC111226/MGC88175/P0/PRLP0/RPP0
Gebräuchliche Bezeichnung RPLP0
Gensymbol RPLP0
Spezies Wheat Germ (in vitro)
Rekombinant Recombinant
Proteinmarkierung GST
Expressionssystem wheat germ expression system
Form Liquid
Mehr anzeigen Weniger anzeigen
Berichtigung von Produktinhalten

Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.

Name des Produkts
Abnova™ Human RPLP0 Full-length ORF (AAH08092, 1 a.a. - 317 a.a.) Recombinant Protein with GST-tag at N-terminal >

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.

Vielen Dank, dass Sie uns helfen, unsere Website zu verbessern. Ihr Feedback wurde übermittelt