missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human RNF25 Full-length ORF (NP_071898.2, 1 a.a. - 459 a.a.) Recombinant Protein with GST-tag at N-terminal
Click to view available options
Menge:
10 μg
25 μg
Packungsgröße:
10 Mikrogramm
25 Mikrogramm
Beschreibung
The protein encoded by this gene contains a RING finger motif. The mouse counterpart of this protein has been shown to interact with Rela, the p65 subunit of NF-kappaB (NFKB), and modulate NFKB-mediated transcription activity. The mouse protein also binds ubiquitin-conjugating enzymes (E2s) and is a substrate for E2-dependent ubiquitination. [provided by RefSeq]
Sequence: MAASASAAAGEEDWVLPSEVEVLESIYLDELQVIKGNGRTSPWEIYITLHPATAEDQDSQYVCFTLVLQVPAEYPHEVPQISIRNPRGLSDEQIHTILQVLGHVAKAGLGTAMLYELIEKGKEILTDNNIPHGQCVICLYGFQEKEAFTKTPCYHYFHCHCLARYIQHMEQELKAQGQEQEQERQHATTKQKAVGVQCPVCREPLVYDLASLKAAPEPQQPMELYQPSAESLRQQEERKRLYQRQQERGGIIDLEAERNRYFISLQQPPAPAEPESAVDVSKGSQPPSTLAAELSTSPAVQSTLPPPLPVATQHICEKIPGTRSNQQRLGETQKAMLDPPKPSRGPWRQPERRHPKGGECHAPKGTRDTQELPPPEGPLKEPMDLKPEPHSQGVEGPPQEKGPGSWQGPPPRRTRDCVRWERSKGRTPGSSYPRLPRGQGAYRPGTRRESLGLESKDGS
Spezifikation
Spezifikation
Zugriffsnummer | NP_071898.2 |
Zur Verwendung mit (Anwendung) | Antibody Production, Protein Array, ELISA, Western Blot |
Zusammensetzung | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gen-ID (Entrez) | 64320 |
Molekulargewicht | 77.6kDa |
Name | RNF25 (Human) Recombinant Protein (P01) |
Reinigungsverfahren | Glutathione Sepharose 4 Fast Flow |
Qualitätskontrollen | 12.5% SDS-PAGE Stained with Coomassie Blue. |
Menge | 10 μg |
Immunogen | MAASASAAAGEEDWVLPSEVEVLESIYLDELQVIKGNGRTSPWEIYITLHPATAEDQDSQYVCFTLVLQVPAEYPHEVPQISIRNPRGLSDEQIHTILQVLGHVAKAGLGTAMLYELIEKGKEILTDNNIPHGQCVICLYGFQEKEAFTKTPCYHYFHCHCLARYIQHMEQELKAQGQEQEQERQHATTKQKAVGVQCPVCREPLVYDLASLKAAPEPQQPMELYQPSAESLRQQEERKRLYQRQQERGGIIDLEAERNRYFISLQQPPAPAEPESAVDVSKGSQPPSTLAAELSTSPAVQSTLPPPLPVATQHICEKIPGTRSNQQRLGETQKAMLDPPKPSRGPWRQPERRHPKGGECHAPKGTRDTQELPPPEGPLKEPMDLKPEPHSQGVEGPPQEKGPGSWQGPPPRRTRDCVRWERSKGRTPGSSYPRLPRGQGAYRPGTRRESLGLESKDGS |
Mehr anzeigen |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Abnova™ Human RNF25 Full-length ORF (NP_071898.2, 1 a.a. - 459 a.a.) Recombinant Protein with GST-tag at N-terminal >
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur