missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human RKHD3 Partial ORF (NP_115622, 60 a.a. - 159 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marke: Abnova™ H00084206-Q01.25ug
Additional Details : Gewicht : 0.00010kg
Beschreibung
Sequence: RKKSVNMTECVPVPSSEHVAEIVGRQGCKIKALRAKTNTYIKTPVRGEEPVFVVTGRKEDVAMARREIISAAEHFSMIRASRNKNTALNGAVPGPPNLPGSpezifikation
NP_115622 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
RKKSVNMTECVPVPSSEHVAEIVGRQGCKIKALRAKTNTYIKTPVRGEEPVFVVTGRKEDVAMARREIISAAEHFSMIRASRNKNTALNGAVPGPPNLPG | |
RUO | |
MEX3B | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
84206 | |
RKHD3 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DKFZp434J0617/MEX-3B/MGC117199/RKHD3/RNF195 | |
MEX3B | |
Recombinant | |
wheat germ expression system |